Skip to Content
MilliporeSigma
All Photos(7)

Key Documents

T0254

Sigma-Aldrich

L-Tryptophan

≥98% (HPLC)

Synonym(s):

(S)-2-Amino-3-(3-indolyl)propionic acid, L-α-Amino-3-indolepropionic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H12N2O2
CAS Number:
Molecular Weight:
204.23
Beilstein:
86197
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.26

Product Name

L-Tryptophan, reagent grade, ≥98% (HPLC)

grade

reagent grade

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to off-white

mp

280-285 °C (dec.) (lit.)

solubility

0.5 M HCl: 50 mg/mL

application(s)

cell analysis

SMILES string

N[C@@H](Cc1c[nH]c2ccccc12)C(O)=O

InChI

1S/C11H12N2O2/c12-9(11(14)15)5-7-6-13-10-4-2-1-3-8(7)10/h1-4,6,9,13H,5,12H2,(H,14,15)/t9-/m0/s1

InChI key

QIVBCDIJIAJPQS-VIFPVBQESA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

L-tryptophan has been used for the isolation of bone marrow–derived neutrophils. It has also been used to identify the use of Raman microscopy as a tool to study the network of neurons in pathologically normal retina.

Biochem/physiol Actions

Tryptophan (Trp) is a heterocyclic, essential amino acid associated with growth, reproduction and immunity. Increased availability of tryptophan is necessary for the regulation of mood, cognition and behavior. The uptake of tryptophan by the brain depends on the plasma ratio of Trp to all of the other large neutral amino acids. Higher the ratio, greater is the Trp uptake.

Other Notes

Amino acid precursor of serotonin and melatonin

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effects of tryptophan loading on human cognition, mood, and sleep.
Neuroscience and Biobehavioral Reviews, 34(3) (2010)
Tryptophan Catabolism Restricts IFN-??Expressing Neutrophils and Clostridium difficile Immunopathology
Mohamad El-Zaatari
Journal of Immunology (2014)
Raman microscopy of porcine inner retinal layers from the area centralis
J. Renwick
Molecular Vision (2007)
Amino acids: metabolism, functions, and nutrition.
Wu G
Amino Acids (2009)
Michael Platten et al.
Cancer research, 72(21), 5435-5440 (2012-10-24)
Tryptophan catabolism in cancer is increasingly being recognized as an important microenvironmental factor that suppresses antitumor immune responses. It has been proposed that the essential amino acid tryptophan is catabolized in the tumor tissue by the rate-limiting enzyme indoleamine-2,3-dioxygenase (IDO)

Articles

Amino Acid Codon Wheel for fast RNA translation. Find which amino acid is translated from your RNA sequence quickly and easily.

Amino Acid Codon Wheel for fast RNA translation. Find which amino acid is translated from your RNA sequence quickly and easily.

Amino Acid Codon Wheel for fast RNA translation. Find which amino acid is translated from your RNA sequence quickly and easily.

Amino Acid Codon Wheel for fast RNA translation. Find which amino acid is translated from your RNA sequence quickly and easily.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service