Skip to Content
MilliporeSigma
All Photos(1)

Documents

NIST1575A

Trace elements in pine needles

NIST® SRM® 1575a

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

packaging

pkg of 50 g powder

manufacturer/tradename

NIST®

application(s)

agriculture

format

matrix material

General description

Each unit of SRM 1575A contains approximately 50 g of pine needles which are dried, jet milled, blended, and sterilized through radiation.

SRM 1575A _SDS

SRM 1575A _CoA

Application

NIST1575A Trace elements in pine needles standard reference material (SRM) are intended for use in the  evaluation of techniques used for the analysis of pine needles and other samples that share similar matrix.

Features and Benefits

  • Accompanied by the certified documenting NIST-certified and non-certified values for minor constituents and trace elements expressed as mass fractions on a dry mass basis.  
  • Certified values are measured by cold vapor isotope dilution inductively coupled plasma mass spectrometry. 
  • Certified values are traceable to the International System of Units (SI) and SI-derived units of mass fraction. 
  • The NIST certificate is provided with storage, source, preparation, analysis details, and use instructions.

Other Notes

Certified for the analytes listed below.
Phosphorus (P), Potassium (K), Calcium (Ca), Aluminum (Al), Barium (Ba), Cadmium (Cd), Chlorine (Cl), Copper (Cu), Iron (Fe), Mercury (Hg), Rubidium (Rb), Zinc (Zn)

Reference/Informational value is provided for the following analytes. Aluminum (Al), Arsenic (As), Barium (Ba), Boron (B), Cadmium (Cd), Calcium (Ca), Cerium (Ce), Cesium, Chlorine (Cl), Chromium (Cr), Cobalt (Co), Copper (Cu), Iron (Fe), Lead (Pb), Magnesium (Mg), Manganese (Mn), Mercury (Hg), Nickel (Ni), Phosphorus (P), Potassium (K), Rubidium (Rb), Scandium (Sc), Selenium (Se), Sodium (Na), Zinc (Zn)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service