Skip to Content
MilliporeSigma
All Photos(1)

Documents

901991

Sigma-Aldrich

Dde biotin picolyl azide

Synonym(s):

N-[6-(Azidomethyl)-3-pyridinyl]-5-(4,4-dimethyl-2,6-dioxocyclohexylidene)-25-[(3aS,4S,6aR)-hexahydro-2-oxo-1H-thieno[3,4-d]imidazol-4-yl]-21-oxo-8,11,14,17-tetraoxa-4,20-diazapentacosanamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C38H57N9O9S
CAS Number:
Molecular Weight:
815.98
UNSPSC Code:
12352125
NACRES:
NA.22

form

solid

reaction suitability

reaction type: click chemistry

storage temp.

−20°C

Application

Dde biotin picolyl azide is a unique alkyne-reactive, cleavable biotin probe that allows for release of the captured biomolecules from streptavidin under mild conditions. This reagent contains a biotin moiety linked to a picolyl azide moiety through a spacer arm containing a hydrazine-cleavable Dde moiety. Captured biomolecules can be efficiently released, typically >90% with 2% hydrazine in aqueous media.
Dde biotin picolyl azide incorporates a copper-chelating motif that dramatically accelerates the Cu(I)-catalyzed azide-alkyne cycloaddition (CuAAC) reaction under conditions relevant to biomolecular labeling. Discovery of azides with an internal Cu(I)-chelating motif greatly enhances the utility of the CuAAC reaction for various applications. Some examples are monitoring the dynamic glycan biosynthesis in living organisms; sensitive detection of metabolically-labeled proteins, DNAs, and RNAs; and many other applications where rate acceleration and reduced cell toxicity are highly desirable.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service