Skip to Content
MilliporeSigma

902020

Desthiobiotin-PEG3-Azide

Synonym(s):

(4S,5R)-N-[2-[2-[2-(2-Azidoethoxy)ethoxy]ethoxy]ethyl]-5-methyl-2-oxo-4-imidazolidinehexanamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C18H34N6O5
CAS Number:
Molecular Weight:
414.50
UNSPSC Code:
51171641
NACRES:
NA.22
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

paste

storage temp.

−20°C

SMILES string

[N+](=[N-])=NCCOCCOCCOCCNC(=O)CCCCC[C@@H]1NC(=O)N[C@@H]1C

InChI

1S/C18H34N6O5/c1-15-16(23-18(26)22-15)5-3-2-4-6-17(25)20-7-9-27-11-13-29-14-12-28-10-8-21-24-19/h15-16H,2-14H2,1H3,(H,20,25)(H2,22,23,26)/t15-,16+/m1/s1

InChI key

JIQVKRRASYKCEL-CVEARBPZSA-N

Application

Desthiobiotin is a single-ring, sulfur-free analog of biotin that binds to streptavidin with nearly equal specificity but less affinity than biotin (Kd = 1011 vs. 1015 M, respectively). Consequently, desthiobiotinylated proteins can be eluted readily and specifically from streptavidin affinity resin using mild conditions based on competitive displacement with free biotin. For pull-down experiments with biological samples, this soft-release characteristic of desthiobiotin also helps to minimize co-purification of endogenous biotinylated molecules, which remain bound to streptavidin upon elution of the target protein complexes with free biotin.


Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library