Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

901763

Sigma-Aldrich

PC biotin-azide

Synonym(s):

rel-1-[4-[[22-[(3aR,4R,6aS)-Hexahydro-2-oxo-1H-thieno[3,4-d]imidazol-4-yl]-4,18-dioxo-8,11,14-trioxa-5,17-diazadocos-1-yl]oxy]-5-methoxy-2-nitrophenyl]ethyl N-(3-azidopropyl)carbamate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C35H55N9O12S
CAS Number:
Molecular Weight:
825.93
MDL number:
UNSPSC Code:
12352125
NACRES:
NA.22

form

powder or crystals

availability

available only in USA

storage temp.

−20°C

SMILES string

S1[C@H]([C@H]3NC(=O)N[C@H]3C1)CCCCC(=O)NCCOCCOCCOCCNC(=O)CCCOc2c(cc(c(c2)[N+](=O)[O-])C(OC(=O)NCCCN=[N+]=[N-])C)OC

InChI key

PTNZFIYMARMNDM-MJUSKHNJSA-N

Application

PC biotin-azide is an azide-activated photoclevable biotin probe that allows for reagent-free release of the captured biomolecules from streptavidin. This reagent contains a biotin moiety linked to an azide group through a spacer arm containing a photocleavable moiety. Captured biomolecules can be efficiently photoreleased, typically >90% in 5-25 minutes using an inexpensive, near-UV, low-intensity lamp (e.g. 365 nm lamp at 1-5 mW/cm2).

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Self-react. C

Storage Class Code

5.2 - Organic peroxides and self-reacting hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service