Skip to Content
MilliporeSigma

901765

Dde biotin-azide

Synonym(s):

rel-(3aR,4R,6aS)-N-[19-Azido-15-(4,4-dimethyl-2,6-dioxocyclohexylidene)-3,6,9,12-tetraoxa-16-azanonadec-1-yl]hexahydro-2-oxo-1H-thieno[3,4-d]imidazole-4-pentanamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C32H53N7O8S
CAS Number:
Molecular Weight:
695.87
UNSPSC Code:
12352125
NACRES:
NA.22
MDL number:
Form:
liquid
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

liquid

reaction suitability

reaction type: click chemistry

storage temp.

−20°C (−15°C to −25°C)

SMILES string

S1[C@H]([C@H]3NC(=O)N[C@H]3C1)CCCCC(=O)NCCOCCOCCOCCOCCC(=C2C(=O)CC(CC2=O)(C)C)NCCCN=[N+]=[N-]

InChI

1S/C32H53N7O8S/c1-32(2)20-25(40)29(26(41)21-32)23(34-9-5-10-36-39-33)8-12-44-14-16-46-18-19-47-17-15-45-13-11-35-28(42)7-4-3-6-27-30-24(22-48-27)37-31(43)38-30/h24,27,30,34H,3-22H2,1-2H3,(H,35,42)(H2,37,38,43)/t24-,27-,30-/m0/s1

InChI key

HUAWNOKWZYCBQA-LFERIPGTSA-N

Application

Extraordinary strength of the streptavidin-biotin interaction allows for efficient capturing of even highly dilute targets; however, it makes recovery of proteins from affinity resins challenging. This reagent contains a biotin moiety linked to an azide moiety through a spacer arm containing a hydrazine-cleavable Dde linker. Typically >90% of captured biomolecules can be efficiently released.


Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library