Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

81101

Supelco

Triethylamine

suitable for HPLC, LiChropur, ≥99.5% (GC)

Synonym(s):

N,N-Diethylethanamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)3N
CAS Number:
Molecular Weight:
101.19
Beilstein:
605283
EC Number:
MDL number:
UNSPSC Code:
41116105
NACRES:
NB.21

vapor density

3.5 (vs air)

Quality Level

vapor pressure

51.75 mmHg ( 20 °C)

product line

LiChropur

Assay

≥99.5% (GC)

form

liquid

autoignition temp.

593 °F

expl. lim.

8 %

technique(s)

HPLC: suitable

refractive index

n20/D 1.401 (lit.)

pH

12.7 (15 °C, 100 g/L)

bp

88.8 °C (lit.)

mp

−115 °C (lit.)

density

0.726 g/mL at 25 °C (lit.)

cation traces

Al: ≤0.2 mg/kg
Ba: ≤0.1 mg/kg
Bi: ≤0.1 mg/kg
Ca: ≤0.5 mg/kg
Cd: ≤0.05 mg/kg
Co: ≤0.02 mg/kg
Cr: ≤0.02 mg/kg
Cu: ≤0.02 mg/kg
Fe: ≤0.1 mg/kg
K: ≤0.5 mg/kg
Li: ≤0.1 mg/kg
Mg: ≤0.1 mg/kg
Mn: ≤0.02 mg/kg
Mo: ≤0.02 mg/kg
Na: ≤0.5 mg/kg
Ni: ≤0.02 mg/kg
Pb: ≤0.1 mg/kg
Sr: ≤0.1 mg/kg
Zn: ≤0.1 mg/kg

absorption

cut-off at 250.0-280.0 nm

SMILES string

CCN(CC)CC

InChI

1S/C6H15N/c1-4-7(5-2)6-3/h4-6H2,1-3H3

InChI key

ZMANZCXQSJIPKH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Triethylamine may be used as a mobile phase in:
  • High-performance liquid chromatography (HPLC) determination of levofloxacin in soft tissue, bone, bile and serum.
  • HPLC determination of phospho- and sphingo-lipids in dairy products.
  • HPLC determination of amoxicillin in premixes.

Packaging

Clear borosilicate glass, high hydrolytic resistance (Type I)

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1A - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

12.2 °F - closed cup

Flash Point(C)

-11 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Analysis of phospho-and sphingolipids in dairy products by a new HPLC method
Rombaut R, et al.
Journal of Dairy Science, 88(2), 482-488 (2005)
An HPLC assay and a microbiological assay to determine levofloxacin in soft tissue, bone, bile and serum
Bottcher S, et al.
Journal of Pharmaceutical and Biomedical Analysis, 25(2), 197-203 (2001)
Rapid determination of amoxicillin in premixes by HPLC
Dousa M and Hosmanova R
Journal of Pharmaceutical and Biomedical Analysis, 37(2), 373-377 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service