Skip to Content
MilliporeSigma
All Photos(1)

Documents

56778

Supelco

Triethylammonium phosphate solution

suitable for HPLC, 0.95-1.05 M

Synonym(s):

Buffer solution 1 M pH 3.0, TEAP

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41116105
NACRES:
NB.21

form

liquid

concentration

0.95-1.05 M

technique(s)

HPLC: suitable

refractive index

n20/D 1.358-1.362

density

1.045-1.600 g/mL at 20 °C

UV absorption

λ: 210 nm Amax: ≤0.30
λ: 220 nm Amax: ≤0.20
λ: 230 nm Amax: ≤0.10
λ: 250 nm Amax: ≤0.04
λ: 500 nm Amax: ≤0.02

storage temp.

2-8°C

SMILES string

OP(O)(O)=O.OP(O)(O)=O.CCN(CC)CC

InChI

1S/C6H15N.H3O4P/c1-4-7(5-2)6-3;1-5(2,3)4/h4-6H2,1-3H3;(H3,1,2,3,4)

InChI key

UNXNGGMLCSMSLH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Triethylammonium phosphate solution may be used as a mobile phase additive for toxicological drug screening by high-performance liquid chromatography (HPLC) and as an elution buffer in HPLC separation of DL-amino acids.

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

HPLC separation of DL-amino acids derivatized withN 2-(5-fluoro-2, 4-dinitrophenyl)-L-amino acid amides
Bruckner H and Keller-Hoehl C
Chromatographia, 30(11-12), 621-629 (1990)
Development of a high-performance liquid chromatography retention index scale for toxicological drug screening
Elliott SP and Hale KA
Journal of Chromatography. B, Biomedical Sciences and Applications, 694(1), 99-114 (1997)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service