Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

56982

Supelco

Potassium phthalate monobasic

suitable for HPLC, LiChropur, 99.5-100.5% (HPLC)

Synonym(s):

KHP, Phthalic acid monopotassium salt, Potassium biphthalate, Potassium hydrogen phthalate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOOCC6H4COOK
CAS Number:
Molecular Weight:
204.22
Beilstein:
3637128
MDL number:
UNSPSC Code:
41116105

Assay

99.5-100.5% (HPLC)

form

solid

quality

LiChropur

technique(s)

HPLC: suitable

loss

≤0.5% loss on drying

pH

3.0-5.0

mp

295-300 °C (dec.) (lit.)

solubility

H2O: 10 applications on large gels, clear, colorless (hot)

λ

10 % in H2O (hot)

UV absorption

λ: 330 nm Amax: ≤0.05
λ: 340 nm Amax: ≤0.03
λ: 350 nm Amax: ≤0.02
λ: 500 nm Amax: ≤0.02

SMILES string

[K+].OC(=O)c1ccccc1C([O-])=O

InChI

1S/C8H6O4.K/c9-7(10)5-3-1-2-4-6(5)8(11)12;/h1-4H,(H,9,10)(H,11,12);/q;+1/p-1

InChI key

IWZKICVEHNUQTL-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

387.1 °F - Pensky-Martens closed cup

Flash Point(C)

197.3 °C - Pensky-Martens closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service