Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

P1088

Sigma-Aldrich

Potassium hydrogen phthalate

BioXtra, ≥99.95%

Synonym(s):

Potassium phthalate monobasic, KHP, Phthalic acid monopotassium salt, Potassium biphthalate, Potassium hydrogen phthalate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOOCC6H4COOK
CAS Number:
Molecular Weight:
204.22
Beilstein:
3637128
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.25
Pricing and availability is not currently available.

product line

BioXtra

Quality Level

Assay

≥99.95%

form

powder or crystals

impurities

<0.005% Insoluble matter
<0.005% Phosphorus (P)

pH

4

mp

295-300 °C (dec.) (lit.)

anion traces

chloride (Cl-): <0.003%
sulfur compounds (as SO42-): ≤0.002%

cation traces

Al: <0.0005%
Ca: <0.005%
Cu: <0.0005%
Fe: <0.0005%
Mg: <0.0005%
NH4+: <0.05%
Na: <0.005%
Pb: <0.001%
Zn: <0.0005%

SMILES string

[K+].OC(=O)c1ccccc1C([O-])=O

InChI

1S/C8H6O4.K/c9-7(10)5-3-1-2-4-6(5)8(11)12;/h1-4H,(H,9,10)(H,11,12);/q;+1/p-1

InChI key

IWZKICVEHNUQTL-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Potassium hydrogen phthalate (KHP) is a semi-organic salt that is a member of the alkali acid phthalate series. KHP crystals are useful in devices because of their electro-optical properties. It usually acts as a model compound due to its well-developed surface pattern on the (010) face that can be easily observed by optical microscopy.

Application

Potassium hydrogen phthalate has been used as a substrate to assess the permeation from the extracellular (EC) to the periplasmic (PP) side through DcaP. It has also been used as one of the polyurethane materials for the characterization of polyester triol (OH).

Features and Benefits

  • Long-term stability
  • Exhibits dielectric properties

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

387.1 °F - Pensky-Martens closed cup

Flash Point(C)

197.3 °C - Pensky-Martens closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Meghan O'Connor et al.
Water research, 156, 23-33 (2019-03-25)
The organic matter present in wastewater effluents (EfOM), is likely to have different properties than the organic matter present in the receiving water. The properties of EfOM will affect the fate of contaminants of emerging concern because EfOM is a
Marta Guembe-García et al.
Polymers, 12(6) (2020-06-04)
We anchored a colourimetric probe, comprising a complex containing copper (Cu(II)) and a dye, to a polymer matrix obtaining film-shaped chemosensors with induced selectivity toward glycine. This sensory material is exploited in the selectivity detection of glycine in complex mixtures
Maria Cristina Fossi et al.
Marine pollution bulletin, 64(11), 2374-2379 (2012-09-12)
Baleen whales are potentially exposed to micro-litter ingestion as a result of their filter-feeding activity. However, the impacts of microplastics on baleen whales are largely unknown. In this case study of the Mediterranean fin whale (Balaenoptera physalus), we explore the
Juanjuan Feng et al.
Journal of chromatography. A, 1268, 16-21 (2012-11-07)
Because of the occurrence of ion exchange between high-ionic-strength solution and anions of polymeric ionic liquids (PILs), PILs based solid-phase microextraction (SPME) fibers were rarely used in direct immersion mode to high-salt-added samples. In this work, a novel double-confined PIL
Hanne Frederiksen et al.
Environmental science & technology, 47(2), 958-967 (2012-12-14)
Urinary phthalate excretion is used as marker of phthalate exposure in epidemiological studies. Here we examine the reliability of urinary phthalate levels in exposure classification by comparing the inter- and intrasubject variation of urinary phthalate metabolite levels. Thirty-three young healthy

Questions

1–4 of 4 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  3. Hi, Is the BioXtra grade equivalent to ACS grade and can we use this reagent for the standardization of NaOH solution as prescribed by the USP pharmacopeia (instead of the ACS grade) ? Thank you in advance.

    1 answer
    1. The BioXtra is not equivalent to the ACS grade. For the standardization of NaOH solutions, it is recommended to use Product 1024000083 which is a certified reference material (CRM) for the standardization of volumetric solutions in alkalimetric titrations, according to Ph. Eur.

      Helpful?

  4. I want to know the expiration date of KHP(P1088)

    1 answer
    1. This product is not assigned an expiration date or recommended retest date. Products with no expiration date or recommended retest date should be routinely inspected by customers to ensure they perform as expected. These products are also subject to a one year warranty from the date of shipment. For more information you may access the "Product Dating Information" document under "ADDITIONAL USEFUL DOCUMENTS ABOUT OUR PRODUCTS" at the bottom of the Quality Services page with this link: https://www.sigmaaldrich.com/life-science/quality-and-regulatory-management/quality-services.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service