Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

52371

Supelco

Hexadecyltrimethylammonium bisulfate

suitable for ion pair chromatography, LiChropur, ≥99.0% (T)

Synonym(s):

Hexadecyltrimethylammonium hydrogen sulfate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)15N(HSO4)(CH3)3
CAS Number:
Molecular Weight:
381.61
Beilstein:
8398798
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NB.21

description

cationic

Quality Level

Assay

≥99.0% (T)

form

solid

quality

LiChropur

technique(s)

ion pair chromatography: suitable

λ

10 % in H2O

UV absorption

λ: 210 nm Amax: 0.15
λ: 220 nm Amax: 0.10
λ: 260 nm Amax: 0.05
λ: 310 nm Amax: 0.02
λ: 500 nm Amax: 0.02

suitability

corresponds to standard for RP gradient test
corresponds to standard for filter test

SMILES string

OS([O-])(=O)=O.CCCCCCCCCCCCCCCC[N+](C)(C)C

InChI

1S/C19H42N.H2O4S/c1-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20(2,3)4;1-5(2,3)4/h5-19H2,1-4H3;(H2,1,2,3,4)/q+1;/p-1

InChI key

UCRJJNVFJGKYQT-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Hexadecyltrimethylammonium bisulfate is an ion pair reagent suitable for anionic separation sorted by carbon chain length.

Application

Hexadecyltrimethylammonium bisulfate may be used as an electrolyte modifier in studying the effect of alkyltrimethylammonium ions on corrosion and electrochemical behavior of Pb-Ca-Sn alloy.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effect of Alkyltrimethylammonium Ions on Corrosion and Electrochemical Behavior of Pb-Ca-Sn Alloy
Kopczynski K, et al.
International Journal of Electrochemical Science, 13, 11058-11073 (2018)
Amit Kumar Mandal et al.
Chemphyschem : a European journal of chemical physics and physical chemistry, 14(4), 788-796 (2012-11-13)
Femtosecond upconversion, single-molecule fluorescence resonance energy transfer (sm-FRET) and fluorescence correlation spectroscopy (FCS) are applied to study the competition between excited-state proton transfer (ESPT) and FRET [to rhodamine 6G (R6G)] of 8-hydroxypyranine-1,3,6-trisulfonate (HPTS) in cetyltrimethylammonium chloride (CTAC) micelles. Pyranine exhibits
Ruggero Angelico et al.
Physical chemistry chemical physics : PCCP, 15(2), 586-595 (2012-11-28)
The aim of the present study was to provide experimental evidence that base pairing, commonly occurring between nucleic bases in more complex supramolecular arrangements, may affect the reaction pathways associated with the alkylation of bases themselves. In pursuit of this
Tanja Mehling et al.
Journal of chromatography. A, 1273, 66-72 (2013-01-01)
Several methods for the description of the retention behavior in micellar liquid chromatography (MLC) were described previously. Thereby, the most common are the linear solvation energy relationships (LSER). However, for the evaluation of the LSER, a number of experimental data
Gurinder Jit Randhawa et al.
Methods in molecular biology (Clifton, N.J.), 958, 139-151 (2012-11-13)
The adoption rate and global area under cultivation of genetically modified (GM) crops is dramatically increasing in recent past. GM cotton has occupied 25.0 million hectares (mha) comprising 15.6% of the global area under GM cultivation. Bt cotton, expressing delta-endotoxins

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service