Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

52366

Sigma-Aldrich

Hexadecyltrimethylammonium chloride

≥98.0% (NT)

Synonym(s):

Cetyltrimethylammonium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)15N(Cl)(CH3)3
CAS Number:
Molecular Weight:
320.00
Beilstein:
3657974
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

description

cationic

Quality Level

Assay

≥98.0% (NT)

mol wt

320.00 g/mol

mp

232-237 °C

SMILES string

[Cl-].CCCCCCCCCCCCCCCC[N+](C)(C)C

InChI

1S/C19H42N.ClH/c1-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20(2,3)4;/h5-19H2,1-4H3;1H/q+1;/p-1

InChI key

WOWHHFRSBJGXCM-UHFFFAOYSA-M

Application

Hexadecyltrimethylammonium chloride (CTAC) is a cationic surfactant that can be used:
  • For the preparation of Polymethylsilsesquioxane aerogels (PMSQ) from methyltrimethoxysilane (MTMS) by hydrolysis and polycondensation reaction.
  • To determine iodine and iodide in commercial gargle products by luminol chemiluminescence.
  • As a template along with cetyltrimethylammonium bromide (CTAB) to prepare rod-like mesoporous silica from sodium silicate as a precursor.
  • As a catalyst in the synthesis of 2-amino-2-chromenes from aldehyde, malononitrile, and phenol by one-pot three-component reaction.

Hexadecyltrimethylammonium chloride can be used as a precursor to synthesize MCM-41 (Mobil Oil, Composition of Matter No. 41), a uniform mesoporous material at room temperature by a two-step sol-gel process.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Corr. 1C

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of rod-like mesoporous silica using mixed surfactants of cetyltrimethylammonium bromide and cetyltrimethylammonium chloride as templates
Han S, et al.
Materials Letters, 57(29), 4520-4524 (2003)
Structure and properties of polymethylsilsesquioxane aerogels synthesized with surfactant n-hexadecyltrimethylammonium chloride
Hayase G, et al.
Microporous and Mesoporous Materials : The Official Journal of the International Zeolite Association, 158, 247-252 (2012)
Catalytic epoxidation of unsaturated alcohols on Ti-MCM-41.
Berlini C, et al.
Catalysis Today, 60(3-4), 219-225 (2000)
Chemiluminescence determination of iodide and/or iodine using a luminol-hexadecyltrimethylammonium chloride reversed micelle system following on-line oxidation and extraction
Fujiwara T, et al.
Analyst, 125(4), 759-763 (2000)
Tharindu A Ranathunge et al.
Nanomaterials (Basel, Switzerland), 9(2) (2019-02-10)
Doxorubicin (DOX) is an anticancer drug commonly used in treating cancer; however, it has severe cytotoxicity effects. To overcome both the adverse effects of the drug and mineral deficiency (i.e., hypomagnesemia) experienced by cancer patients, we have developed magnesium oxide

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service