MilliporeSigma
Alkylation of complementary ribonucleotides in nanoreactors.
Physical chemistry chemical physics : PCCP (2012-11-28)
Ruggero Angelico, Ilario Losito, Francesca Cuomo, Andrea Ceglie, Francesco Palmisano
ABSTRACT
The aim of the present study was to provide experimental evidence that base pairing, commonly occurring between nucleic bases in more complex supramolecular arrangements, may affect the reaction pathways associated with the alkylation of bases themselves. In pursuit of this aim, dilute aqueous solutions of Cytidine- (CMP) and Guanosine-Mono-Phosphate (GMP) as single reactants or in an equimolar mixture were treated with the electrophilic alkylating agent 1,2-Dodecyl-Epoxide (DE), which was preventively dispersed into micellar solutions prepared with the cationic surfactant hexadecyltrimethylammonium bromide (CTAB). In the early stage of the reaction, CTAB micelles acted as micro-heterogeneous nanoreactors, but as the reaction progressed the systems evolved toward the formation of polydisperse aggregates, whose size and surface-charge properties were monitored as a function of reaction time. From mass spectrometry analyses, it was found that the deamination of cytosine, a side reaction related to the alkylation of the amino group of CMP, was reduced when both the complementary ribonucleotides were present in the same reaction mixture. The involvement of specific sites able to establish C:G interactions (possibly via H-bonding or π-π stacking) could explain the reduced reactivity occurring at the level of some of the nucleophilic centers responsible for molecular recognition.
Sigma-Aldrich
Cetyltrimethylammonium hydrogensulfate, 99%
Sigma-Aldrich
Hexadecyltrimethylammonium chloride, ≥98.0% (NT)
Sigma-Aldrich
Cetyltrimethylammonium chloride solution, 25 wt. % in H2O
Sigma-Aldrich
Hexadecyltrimethylammonium hydroxide solution, ~25% in methanol (T)
Sigma-Aldrich
Hexadecyltrimethylammonium hydroxide solution, 10 wt. % in H2O
Sigma-Aldrich
Cytidine 5′-monophosphate, Sigma Grade, ≥99% (HPLC), synthetic, powder
Supelco
Hexadecyltrimethylammonium bromide, analytical standard
Supelco
Hexadecyltrimethylammonium bisulfate, suitable for ion pair chromatography, LiChropur, ≥99.0% (T)
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, ≥96.0% (AT)
Supelco
Hexadecyltrimethylammonium bromide, suitable for ion pair chromatography, LiChropur
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, for molecular biology, ≥99%
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioXtra, ≥99%
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioUltra, for molecular biology, ≥99.0% (AT)
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, ≥98%