Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

77626

Supelco

N-tert-Butyldimethylsilyl-N-methyltrifluoroacetamide

for GC derivatization, LiChropur, ≥98.0% (GC)

Synonym(s):

MTBSTFA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3CON(CH3)Si(CH3)2C(CH3)3
CAS Number:
Molecular Weight:
241.33
Beilstein:
3606546
MDL number:
UNSPSC Code:
12352111
NACRES:
NA.22

grade

for GC derivatization

Assay

≥98.0% (GC)

form

liquid

quality

LiChropur

reaction suitability

reagent type: derivatization reagent
reaction type: Silylations

technique(s)

gas chromatography (GC): suitable

refractive index

n20/D 1.402 (lit.)

bp

172-175 °C (lit.)

density

1.036 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CN(C(=O)C(F)(F)F)[Si](C)(C)C(C)(C)C

InChI

1S/C9H18F3NOSi/c1-8(2,3)15(5,6)13(4)7(14)9(10,11)12/h1-6H3

InChI key

QRKUHYFDBWGLHJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

N-tert-Butyldimethylsilyl-N-methyltrifluoroacetamide is a silylating reagent which replaces the active hydrogen with tert-Butyldimethylsilyl group. The tert-Butyldimethylsilyl derivatives are found to be more resistant to hydrolysis and stable compared to trimethylsilyl (TMS) derivatives. This reagent is suitable for GC-MS analysis since it produces mass spectra, which can be easily interpreted. It is widely used for the silylation of different alcohols, thiols, phenols, carboxylic acids, amines, and amides.

Application

N-tert-Butyldimethylsilyl-N-methyltrifluoroacetamide may be used as a derivatizing reagent for the determination of methionine sulfoxide and methionine in blood plasma and chlorpyrifos, diazinon and their major metabolites in sludge and sludge-fertilized agricultural soils using gas-chromatography coupled to mass spectrometry technique.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

113.0 °F - closed cup

Flash Point(C)

45 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Advanced Gas Chromatography: Progress in Agricultural, Biomedical and Industrial Applications (2012)
Ryuichi Mashima et al.
Analytical biochemistry, 313(1), 28-33 (2003-02-11)
Methionine sulfoxide is an oxidation product of methionine with reactive oxygen species via 2-electron-dependent mechanism. Such oxidants can be generated from activated neutrophils; therefore, methionine sulfoxide can be regarded as a biomarker of oxidative stress in vivo. We describe here
Two stage derivatization with N-(tert.-butyldimethylsilyl)-N-methyl-trifluoroacetamide (MTBSTFA) and N-methyl-bis-(trifluoroacetamide)(MBTFA) for the gas-chromatographic analysis of OH-, SH-and NH-compounds
Schoene K, et al.
Fresenius Journal of Analytical Chemistry, 348(5-6), 364-370 (1994)
Highly selective sample preparation and gas chromatographic?mass spectrometric analysis of chlorpyrifos, diazinon and their major metabolites in sludge and sludge-fertilized agricultural soils
Diaz-Cruz SM and Barcelo D
Journal of Chromatography A, 1132(1-2), 21-27 (2006)

Articles

Results of a study involving the ability few Fluka silylating reagents to form GC-MS-compatible trimethylsilylmethyl derivatives of NSAIDs

Results of a study involving the ability few Fluka silylating reagents to form GC-MS-compatible trimethylsilylmethyl derivatives of NSAIDs

Results of a study involving the ability few Fluka silylating reagents to form GC-MS-compatible trimethylsilylmethyl derivatives of NSAIDs

Results of a study involving the ability few Fluka silylating reagents to form GC-MS-compatible trimethylsilylmethyl derivatives of NSAIDs

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service