Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

30050

Sigma-Aldrich

Cystamine dihydrochloride

purum, ≥98.0% (AT)

Synonym(s):

2,2′-Diaminodiethyl disulfide dihydrochloride, 2,2′-Dithiobis(ethylamine) dihydrochloride, Decarboxycystine dihydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2CH2CH2SSCH2CH2NH2 · 2HCl
CAS Number:
Molecular Weight:
225.20
Beilstein:
3616850
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

purum

Quality Level

Assay

≥98.0% (AT)

form

powder

mp

217-220 °C (dec.) (lit.)
~220 °C (dec.)

solubility

water: soluble 1 g/10 mL, clear, colorless

functional group

amine
disulfide

SMILES string

Cl[H].Cl[H].NCCSSCCN

InChI

1S/C4H12N2S2.2ClH/c5-1-3-7-8-4-2-6;;/h1-6H2;2*1H

InChI key

YUFRRMZSSPQMOS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Cystamine dihydrochloride was used:
  • as sulfhydryl modifying reagent
  • in synthesis of monoclonal anti-prostate-specific membrane antigen antibody and A9 RNA aptamers-conjugated popcorn shape gold nanoparticles
  • as internal standard in determination of total homocysteine
  • in modification of gold nanorod surface by amine groups

Other Notes

Sulfhydryl modifying reagent

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anant K Singh et al.
ACS nano, 3(7), 1906-1912 (2009-07-04)
The presence of E. coli in foodstuffs and drinking water is a chronic worldwide problem. The worldwide food production industry is worth about U.S. $578 billion, and the demand for biosensors to detect pathogens and pollutants in foodstuffs is growing
Nan-Fu Chiu et al.
International journal of nanomedicine, 14, 4833-4847 (2019-07-17)
The use of functionalized graphene oxide (fGO) has led to a new trend in the sensor field, owing to its high sensitivity with regards to sensing characteristics and easy synthesis procedures. In this study, we developed an ultra-sensitive carboxyl-graphene oxide
L J Riddell et al.
The American journal of clinical nutrition, 71(6), 1448-1454 (2000-06-06)
Elevated plasma total homocysteine (tHcy) concentrations are associated with increased risk of vascular disease, and there is a strong inverse association between dietary and blood folate and blood tHcy concentrations. Increased folate consumption may lower the risk of tHcy-mediated cardiovascular
Wentong Lu et al.
Journal of the American Chemical Society, 132(51), 18103-18114 (2010-12-07)
Prostate cancer is the second leading cause of cancer-related death among the American male population, and the cost of treating prostate cancer patients is about $10 billion/year in the United States. Current treatments are mostly ineffective against advanced-stage prostate cancer
J E Bodwell et al.
Biochemistry, 23(7), 1392-1398 (1984-03-27)
Glucocorticoid -receptor complexes from intact rat thymus cells incubated with [3H]dexamethasone at 0 degree C are in the nonactivated form and do not bind to DNA-cellulose. Upon being warmed, they are transformed to activated complexes that bind to DNA-cellulose at

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service