Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

30080

Sigma-Aldrich

Cysteamine hydrochloride

≥97.0% (RT)

Synonym(s):

β-Mercapto­ethylamine hydrochloride, 2-Amino­ethane­thiol hydrochloride, 2-Mercapto­ethyl­amine hydrochloride, Decarboxy­cysteine hydrochloride, Thio­ethanol­amine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HSCH2CH2NH2 · HCl
CAS Number:
Molecular Weight:
113.61
Beilstein:
3590083
EC Number:
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51

Quality Level

Assay

≥97.0% (RT)

form

crystals

mp

64-70 °C
67-71 °C

storage temp.

2-8°C

SMILES string

Cl[H].NCCS

InChI

1S/C2H7NS.ClH/c3-1-2-4;/h4H,1-3H2;1H

InChI key

OGMADIBCHLQMIP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Cysteamine hydrochloride has been used in the in-situ generation of chiral cysteamine-capped cadmium sulfide quantum dots (CA-CdS QDs) for the sensing of Cd2+ and S2-.

Biochem/physiol Actions

Cysteamine hydrochloride is derived from the degradation of coenzyme A endogenously. Its concentration in plasma is low. It is commonly used as a drug for the orphan disease cystinosis, a lysosomal storage disorder, in which cysteamine decreases intralysosomal cystine accumulation.
Cysteamine is an aminothiol that can reduce important oxidized disulfide molecules such as cystine to yield cysteine. Cysteamine is used in a wide range of applications from regulation of gene expression, to depletion of somatostatin, to coating of nanoparticles.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A circular dichroism sensor for selective detection of Cd2+ and S2- based on the in-situ generation of chiral CdS quantum dots
Sianglam P, et al.
Spectrochemical Analysis by Atomic Absorption and Emission, 183, 408-416 (2017)
Mengxiao Yu et al.
Journal of the American Chemical Society, 133(29), 11014-11017 (2011-07-01)
pH regulates many cellular processes and is also an indicator of disease progression. Therefore, pH-responsive materials often serve as either tools in the fundamental understanding of cell biology or medicine for disease diagnosis and therapy. While gold nanoparticles have broad
Nida Zaman Khan et al.
ACS omega, 6(40), 25926-25939 (2021-10-19)
Heparin-induced thrombocytopenia (HIT) antibodies (Abs) can mediate and activate blood cells, forming blood clots. To detect HIT Abs, immunological assays with high sensitivity (≥95%) and fast response are widely used, but only about 50% of these tests are accurate as
Martine Besouw et al.
Drug discovery today, 18(15-16), 785-792 (2013-02-19)
Cysteamine is an amino thiol with the chemical formula HSCH2CH2NH2. Endogenously, cysteamine is derived from coenzyme A degradation, although its plasma concentrations are low. Most experience with cysteamine as a drug originates from the field of the orphan disease cystinosis
Jiying Pei et al.
Analytica chimica acta, 757, 63-68 (2012-12-05)
High-quality cysteamine-coated CdTe quantum dots (CA-CdTe QDs) were successfully synthesized in aqueous phase by a facile one-pot method. Through hydroxylamine hydrochloride-promoted kinetic growth strategy, water-soluble CA-CdTe QDs could be obtained conveniently in a conical flask by a stepwise addition of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service