Skip to Content
MilliporeSigma
All Photos(3)

Documents

914592

Sigma-Aldrich

Fmoc-Cystamine HCl

≥95%

Synonym(s):

(9H-fluoren-9-yl)methyl 2-((2-aminoethyl)disulfanyl)ethylcarbamate hydrochloride, 2-((9-Fluorenylmethyloxycarbonylamino)ethyl)disulfanyl-(2-aminoethane) hydrochloride, Fmoc-SS-NH2 HCl, Protected cleavable linker

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H23ClN2O2S2
Molecular Weight:
410.98
UNSPSC Code:
12352116
NACRES:
NA.22

Quality Level

Assay

≥95%

form

powder

mp

135 °C (decomp.)

functional group

Fmoc

storage temp.

2-8°C

Application

Fmoc-Cystamine HCl is a valuable building block for the incorporation of disulfides. It possesses two terminal amino functional groups, one with a protecting group. This Fmoc-protected cleavable linker can also be used in the synthesis of chemical biology tools for cleavage under reducing conditions.

related product

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service