Skip to Content
MilliporeSigma
All Photos(1)

Documents

27393

Supelco

1,4-Cineole

analytical standard

Synonym(s):

1-Isopropyl-4-methyl-7-oxabicyclo[2.2.1]heptane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H18O
CAS Number:
Molecular Weight:
154.25
Beilstein:
104974
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.5% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.445 (lit.)
n20/D 1.445

bp

65 °C/16 mmHg (lit.)

mp

−46 °C (lit.)

density

0.887 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

CC(C)C12CCC(C)(CC1)O2

InChI

1S/C10H18O/c1-8(2)10-6-4-9(3,11-10)5-7-10/h8H,4-7H2,1-3H3/t9-,10+

InChI key

RFFOTVCVTJUTAD-AOOOYVTPSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

116.6 °F - closed cup

Flash Point(C)

47 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

L Ji et al.
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica, 22(8), 489-492 (1997-08-01)
The essential oils from the dried stems of Ephedra sinica, E. intermedia and E. equisetina were analyzed by GC-MS qualitatively and GC quantitatively. One hundred and twenty-seven constituents were identified, l-alpha-terpineol (31.64%) in E. sinica, 1,4-cineole (12.80%) in E. intermedia
Y Asakawa et al.
Xenobiotica; the fate of foreign compounds in biological systems, 18(10), 1129-1134 (1988-10-01)
1. The metabolism of 1,4-cineole, a monoterpene ether, was studied in the rabbit. 2. Four neutral and one acidic metabolites were isolated from the urine and shown to be 9-hydroxy-1,4-cineole, 3,8-dihydroxy-1,4-cineole, 8,9-dihydroxy-1,4-cineole, 1,4-cineole-8-en-9-ol and 1,4-cineole-9-carboxylic acid.
Abdul Ahad et al.
Current drug delivery, 8(2), 213-224 (2011-01-18)
The purpose of this study was to investigate the effectiveness and mechanism(s) of percutaneous absorption of propranolol hydrochloride (PHCL) across rat and human cadaver skin using seven novel terpenes with reference to marker terpene 1,8-cineole. In-vitro skin permeation studies were
Patrícia Bezerra Gomes et al.
Pharmacology, biochemistry, and behavior, 96(3), 287-293 (2010-07-31)
Recent studies have shown that some monoterpenes exert anxiolytic- and depressant-like actions, however, these effects from monoterpene 1,4-cineole are still unknown. This work aimed to study the effects of 1,4-cineole in classic animal models for depression- and anxiety-like behavior, specifically
W Liu et al.
Applied and environmental microbiology, 59(11), 3889-3893 (1993-11-01)
A cytochrome P-450-dependent monooxygenase system that catalyzes the stereospecific hydroxylation of the monoterpene substrate 1,4-cineole was demonstrated in cell-free preparations of Bacillus cereus UI-1477. 1,4-Cineole hydroxylations were catalyzed by a 100,000 x g (1-h)-centrifuging soluble, hexane-inducible enzyme that activated and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service