Skip to Content
MilliporeSigma

06941

(N,N-Dimethylmyristylammonio)acetate

≥97.0% (HPLC)

Synonym(s):

(N,N-Dimethyltetradecylammonio)acetate, N,N-Dimethyl-N-myristylaminoacetic acid betaine, N,N-Dimethyl-N-tetradecylbetaine, N,N-Dimethyl-N-tetradecylglycine, Myristyl betaine, Myristyldimethylbetaine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
CH3(CH2)13N+(CH3)2CH2COO-
CAS Number:
Molecular Weight:
299.49
NACRES:
NA.25
PubChem Substance ID:
UNSPSC Code:
12161900
EC Number:
220-006-9
MDL number:
Beilstein/REAXYS Number:
4141460
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥97.0% (HPLC)

mol wt

299.49 g/mol

technique(s)

HPLC: suitable

pH

5.0-8.0

solubility

H2O: 1%, clear, colorless

UV absorption

λ: 260 nm Amax: ≤0.1, λ: 280 nm Amax: ≤0.08

InChI

1S/C18H37NO2/c1-4-5-6-7-8-9-10-11-12-13-14-15-16-19(2,3)17-18(20)21/h4-17H2,1-3H3

InChI key

KKMIHKCGXQMFEU-UHFFFAOYSA-N

Analysis Note

residue (filter test): no residue, luminescence: corresponds to requirements


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library