Skip to Content
MilliporeSigma
1,4-Cineole analytical standard

1,4-Cineole

1,4-Cineole analytical standard
Synonym(s):
1-Isopropyl-4-methyl-7-oxabicyclo[2.2.1]heptane
Empirical Formula (Hill Notation):
C10H18O
CAS Number:
Molecular Weight:
154.25
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Agency
Format
Chemical Composition
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing