MilliporeSigma
Biotransformation of 1,4-cineole, a monoterpene ether.
Xenobiotica; the fate of foreign compounds in biological systems (1988-10-01)
Y Asakawa, M Toyota, T Ishida
ABSTRACT
1. The metabolism of 1,4-cineole, a monoterpene ether, was studied in the rabbit. 2. Four neutral and one acidic metabolites were isolated from the urine and shown to be 9-hydroxy-1,4-cineole, 3,8-dihydroxy-1,4-cineole, 8,9-dihydroxy-1,4-cineole, 1,4-cineole-8-en-9-ol and 1,4-cineole-9-carboxylic acid.
Sigma-Aldrich
1,4-Cineole, technical, mixture of isomers, ≥85% (GC)
Supelco
1,4-Cineole, analytical standard