11229
Ammonium sulfite solution
technical, 34%
Synonym(s):
Diammonium sulfite, Diammonium sulfonate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
grade
technical
Quality Level
concentration
34%
~34% in H2O
SMILES string
N.N.OS(O)=O
InChI
1S/2H3N.H2O3S/c;;1-4(2)3/h2*1H3;(H2,1,2,3)
InChI key
PQUCIEFHOVEZAU-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Ammonium sulfite is an ammonium salt of sulfurous acid and a byproduct of the flue gas desulfuration process by ammonia. Ammonium sulfite can be oxidized into ammonium sulfate, which can be used as a fertilizer.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2
Supplementary Hazards
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Nucleic acids symposium series (2004), (53)(53), 217-218 (2009-09-15)
Bisulfite modification is a principal tool for analyzing DNA methylation, the methyl substitution at position 5 of cytosine residues. Hypermethylation is known to cause silencing of genes, which may result in cell function failures. DNA methylation analysis is therefore a
Oxidation of ammonium sulfite in aqueous solutions using ozone technology
Journal of Physics. Conference Series, 418(1), 012130-012130 (2013)
Generation, behavior, and toxicity of ammonium sulfite aerosols.
Journal of the Air Pollution Control Association, 36(1), 55-59 (1986-01-01)
Nucleic acids symposium series (2004), (51)(51), 47-48 (2007-11-22)
Methylation at position 5 of cytosine in DNA plays a major role in epigenetic gene control. The methylation analysis can be performed by bisulfite genomic sequencing. Conventional procedures in this analysis include a treatment of single stranded DNA with 3-5
Huan jing ke xue= Huanjing kexue, 22(4), 117-119 (2001-09-25)
The experiment results showed that the best technology of removing the nitrogen was anoxic time to aerobic time of 1:1.5, a cycle of 8 hours; the SRT should be more than 12 d and the loading of NH3-N should be
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service