MilliporeSigma
Progress in the bisulfite modification of nucleic acids.
Nucleic acids symposium series (2004) (2009-09-15)
Hikoya Hayatsu, Kazuo Negishi, Yusuke Wataya
ABSTRACT
Bisulfite modification is a principal tool for analyzing DNA methylation, the methyl substitution at position 5 of cytosine residues. Hypermethylation is known to cause silencing of genes, which may result in cell function failures. DNA methylation analysis is therefore a focus of attention in various fields of biological sciences, including even clinical practices for treatment of cancer patients. In 2004, we reported that the bisulfite modification of DNA necessary in this analysis can be speeded up significantly by using a high concentration ammonium bisulfite solution (10 M), in place of traditional sodium bisulfite solution of 5 M concentration. Evaluations on this newer protocol have now come out from several laboratories, showing that this quick process can yield results with greater accuracy compared to those obtainable with widely-practiced low-concentration methods. Another aspect reported here is a study on the desulfonation of uracil-bisulfite adduct to form uracil, the last step of the bisulfite-conversion of cytosine to uracil. Kinetic measurements for the desulfonation of uridine-bisulfite adduct at a near-neutral pH region are described.
Sigma-Aldrich
Ammonium sulfite solution, technical, 34%