Skip to Content
MilliporeSigma
All Photos(1)

Documents

W205303

Sigma-Aldrich

Ammonium sulfide solution

20 wt. % in H2O

Synonym(s):

Ammonium monosulfide, Diammonium sulphide, Diazanium sulfide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(NH4)2S
CAS Number:
Molecular Weight:
68.14
FEMA Number:
2053
Council of Europe no.:
482
MDL number:
UNSPSC Code:
12164502
eCl@ss:
38030205
PubChem Substance ID:
Flavis number:
16.002
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

Halal
Kosher

reg. compliance

FDA 21 CFR 172.515

concentration

20 wt. % in H2O

refractive index

n20/D 1.388

density

0.997 g/mL at 25 °C

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

sulfurous

SMILES string

N.N.S

InChI

1S/2H3N.H2S/h2*1H3;1H2

InChI key

UXXGWUQNRMPNKH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ammonium sulfide is an organosulfur compound, mainly used as a flavor ingredient due to its characteristic sulfurous odor. The fried bacon aroma produced during the reaction of ammonium sulfide with isovaleraldehyde is due to 2,4,6-triisobutyl-4H-1,3,5-dithiazine and 3,5-diisobutyl-1,2,4-trithiolane formed during the reaction.

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

90.0 °F - closed cup

Flash Point(C)

32.2 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Burdock GA
Encyclopedia of Food and Color Additives, 1, 146-146 (1997)
Characterization of bacon odor and other flavor components from the reaction of isovaleraldehyde and ammonium sulfide.
Shu CK, et al.
Journal of Agricultural and Food Chemistry, 33(1), 130-132 (1985)
Burdock GA
Encyclopedia of Food and Color Additives, 1, 146-146 (1997)
[Photographic models in the study and valuation of the silver reactions in histology. II. Optical density variation in enlarging exposure scale under the action of the ammonium sulphide on silver (author's transl)].
M Vialli et al.
Mikroskopie, 36(3-4), 117-127 (1980-06-01)
Shuxia Xu et al.
Luminescence : the journal of biological and chemical luminescence, 25(4), 294-299 (2009-07-07)
In the present work, Fe(3)O(4)-carbon nanotubes (CNTs) composite was explored as a sensing material candidate for ammonium sulfide. Intense chemiluminescence emission can be observed during the catalytic oxidation of ammonium sulfide on the surface of Fe(3)O(4)-CNTs composite. Based on this

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service