Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

06411

Supelco

Amberlite IRA-67 Ion Exchange Resin

free base, 25-35 mesh

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41116105
NACRES:
SB.52

product name

Amberlite IRA-67 free base, free base

Quality Level

form

beads

moisture

~60%

technique(s)

LPLC: suitable

loss

~55% loss on drying, 110 °C

matrix

Acrylic (gel)

matrix active group

tertiary amine

particle size

500-750 μm

capacity

1.6 meq/mL by wetted bed volume

separation technique

anion exchange

InChI

1S/C12H18O3.C10H10.C8H16N2O/c1-3-5-7-13-9-11-15-12-10-14-8-6-4-2;1-3-9-7-5-6-8-10(9)4-2;1-4-8(11)9-6-5-7-10(2)3/h3-4,9-12H,1-2,5-8H2;3-8H,1-2H2;4H,1,5-7H2,2-3H3,(H,9,11)/b11-9+,12-10+;;

InChI key

ZGXNLYRRYYOMNG-JDDKLYJPSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Weakly basic anion exchange resin with an unusually high capacity for large organics. Common applications include: deacidification, deionization of process liquors, isolation of acidic natural products, purification of novabiacin and cephalosporins, separation of neutral and acidic amino acids below pH 10, removal of heparinic acid at low pH, concentration of citric acid, and demineralization of cheese whey.

Other Notes

Previously known as IRA-68.

Legal Information

Amberlite is a trademark of DuPont de Nemours, Inc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service