Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

01380

Supelco

Ethylene carbonate

Standard for quantitative NMR, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

1,3-Dioxolan-2-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H4O3
CAS Number:
Molecular Weight:
88.06
Beilstein:
106249
EC Number:
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:
NACRES:
NA.24

grade

Standard for quantitative NMR
TraceCERT®

Quality Level

vapor density

3.04 (vs air)

vapor pressure

0.02 mmHg ( 36.4 °C)

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

technique(s)

qNMR: suitable ((standard))
qNMR: suitable

bp

243-244 °C/740 mmHg (lit.)

mp

35-38 °C (lit.)

density

1.321 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care
pharmaceutical

storage temp.

2-8°C

SMILES string

O=C1OCCO1

InChI

1S/C3H4O3/c4-3-5-1-2-6-3/h1-2H2

InChI key

KMTRUDSVKNLOMY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food. Find all available reference materials for compounds listed in 10/2011 here

Application


  • Enhancement of chiral synthesis: Ethylene carbonate is utilized as a solvent in iridium-catalyzed asymmetric synthesis processes, improving the production of chiral N-substituted 2-pyridones, which are valuable in pharmaceutical applications (Wang et al., 2024).

  • Corrosion behavior in battery electrolytes: Studies on the corrosion behavior of aluminum current collectors in LiFSI electrolytes highlight the role of ethylene carbonate in enhancing the stability and efficiency of battery electrolytes (Yuan et al., 2024).

  • High-safety quasi-solid-state batteries: Research involving in situ polymerization facilitated by ethylene carbonate leads to the development of practical high-safety quasi-solid-state batteries, enhancing energy storage technologies (Rui et al., 2024).

  • Voltage oscillation in sodium-ion batteries: Ethylene carbonate′s properties are critical in understanding and managing voltage oscillation in sodium-ion batteries, particularly in extreme conditions such as operating at -40 °C (Que et al., 2024).

  • Long-life cycling of lithium-metal batteries: The use of ethylene carbonate in in situ polymerized quasi-solid electrolytes compounded with ionic liquid contributes to the long-life cycling and operational stability of high-voltage (4.45 V) lithium-metal batteries (Ma et al., 2024).

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - STOT RE 2 Oral

Target Organs

Kidney

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

289.4 °F - closed cup

Flash Point(C)

143 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Method development in quantitative NMR towards metrologically traceable organic certified reference materials used as 31P qNMR standards.
Weber, M., Hellriegel, C., Rueck, A., Wuethrich, J., Jenks, P., and Obkircher, M.
Analytical and Bioanalytical Chemistry (2014)
Using high-performance quantitative NMR (HP-qNMR) for certifying traceable and highly accurate purity values of organic reference materials with uncertainties <0.1 %
Weber, M., Hellriegel, C., Rueck, A., Sauermoser, R., and Wuethrich, J.
Accreditation and Quality Assurance, 18, 91-98 (2013)
Michael Weber et al.
Journal of pharmaceutical and biomedical analysis, 93, 102-110 (2013-11-05)
Quantitative NMR spectroscopy (qNMR) is gaining interest across both analytical and industrial research applications and has become an essential tool for the content assignment and quantitative determination of impurities. The key benefits of using qNMR as measurement method for the

Articles

NMR shift charts assist in identifying impurities in deuterated solvents, aiding accurate chemical analysis.

NMR shift charts assist in identifying impurities in deuterated solvents, aiding accurate chemical analysis.

NMR shift charts assist in identifying impurities in deuterated solvents, aiding accurate chemical analysis.

NMR shift charts assist in identifying impurities in deuterated solvents, aiding accurate chemical analysis.

See All

Related Content

ChemisTwin automates NMR result interpretation, saving time with structure confirmation and quantitative NMR measurements using electronic Reference Materials.

ChemisTwin automates NMR result interpretation, saving time with structure confirmation and quantitative NMR measurements using electronic Reference Materials.

ChemisTwin automates NMR result interpretation, saving time with structure confirmation and quantitative NMR measurements using electronic Reference Materials.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service