Skip to Content
MilliporeSigma
All Photos(2)

Documents

809950

Sigma-Aldrich

Ethylene carbonate

battery grade, ≥99%, acid <10 ppm, H2O <10 ppm

Synonym(s):

EC, 1,3-Dioxolan-2-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H4O3
CAS Number:
Molecular Weight:
88.06
Beilstein:
106249
EC Number:
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.23

grade

battery grade

Quality Level

vapor density

3.04 (vs air)

vapor pressure

0.02 mmHg ( 36.4 °C)

Assay

≥99%

form

solid

impurities

≤10 ppm H2O
≤10 ppm acid

bp

243-244 °C/740 mmHg (lit.)

mp

35-38 °C (lit.)
36 °C (exp.)

density

1.321 g/mL at 25 °C (lit.)

application(s)

battery manufacturing

SMILES string

O=C1OCCO1

InChI

1S/C3H4O3/c4-3-5-1-2-6-3/h1-2H2

InChI key

KMTRUDSVKNLOMY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Anhydrous ethylene carbonate is a cyclic ester and chemically related to ethylene glycol and dimethyl carbonate. It is a highly polar solvent and is a good solvent for many organic and inorganic compounds; however, at room temperature (25 °C) ethylene carbonate is a transparent crystalline solid, practically odorless and colorless, and somewhat soluble in water. In the liquid state (m.p. 35-37 °C) ethylene carbonate is a colorless, odorless liquid with a high boiling point (243 °C). Anhydrous ethylene carbonate is a solvent that is commonly used in the production of batteries, particularly lithium-ion batteries, and as a solvent in the pharmaceutical and chemical industries, in part because of its very high dielectric constant. Because of its high melting point and moderate viscosity, ethylene carbonate is often blended with other carbonates to yield a solution at room temperature with lower viscosity.

Application

Alkyl carbonates find applications as solvents for lithium ion battery electrolytes and the use of high quality battery grade electrolytes having extremely low water (<10 ppm) and acid (<10 ppm) contents are critical for achieving high electrochemical performance.

Caution

These additives have low water content (less than 100 ppm). Please handle under inert and moisture free environment (glove box). Keep containers tightly closed. Keep away from heat and ignition sources. Store in a cool and dry place. Avoid storing together with oxidizers.

Legal Information

Product of MU Ionic Solutions Corp

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - STOT RE 2 Oral

Target Organs

Kidney

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

289.4 °F - closed cup

Flash Point(C)

143 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Experts discuss challenges and production processes of nickel-rich layered oxide cathode materials in energy storage systems.

Experts discuss challenges and production processes of nickel-rich layered oxide cathode materials in energy storage systems.

Experts discuss challenges and production processes of nickel-rich layered oxide cathode materials in energy storage systems.

Experts discuss challenges and production processes of nickel-rich layered oxide cathode materials in energy storage systems.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service