Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

901593

Sigma-Aldrich

Ethylene carbonate solution

1 M in THF

Synonym(s):

1,3-Dioxolan-2-one, Glycol carbonate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C3H4O3
CAS Number:
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.22

form

liquid

concentration

1 M in THF

refractive index

n/D 1.4095

density

0.9079

SMILES string

O1CCOC1=O

InChI

1S/C3H4O3/c4-3-5-1-2-6-3/h1-2H2

InChI key

KMTRUDSVKNLOMY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Valuable building block or solvent offered as a solution in tetrahydrofuran for more convenient handling.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - STOT RE 2 Oral - STOT SE 3

Target Organs

Kidney, Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

0.5 °F

Flash Point(C)

-17.5 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service