Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

901612

Sigma-Aldrich

Ethylene carbonate solution

1 M in DMF

Synonym(s):

1,3-Dioxolan-2-one, Glycol carbonate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C3H4O3
CAS Number:
UNSPSC Code:
12352200
NACRES:
NA.22

form

liquid

reaction suitability

reagent type: reductant

concentration

1 M in DMF

density

0.984 g/mL

InChI

1S/C3H4O3/c4-3-5-1-2-6-3/h1-2H2

InChI key

KMTRUDSVKNLOMY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Valuable building block or solvent offered as a solution in dimethylformamide for more convenient handling.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Eye Irrit. 2 - Flam. Liq. 3 - Repr. 1B - STOT RE 2 Oral

Target Organs

Kidney

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

137.3 °F

Flash Point(C)

58.5 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service