Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

15-1090

Sigma-Aldrich

Ammonium iron(II) sulfate solution

0.1 M

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
form:
liquid

form

liquid

reaction suitability

reagent type: catalyst
core: iron

availability

available only in Japan

concentration

0.1 M
1/10 N

storage temp.

15-25°C

SMILES string

N.N.[Fe++].OS([O-])(=O)=O.OS([O-])(=O)=O

InChI

1S/Fe.2H3N.2H2O4S/c;;;2*1-5(2,3)4/h;2*1H3;2*(H2,1,2,3,4)/q+2;;;;/p-2

InChI key

IMBKASBLAKCLEM-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service