Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.09864

Supelco

Ammonium iron(II) sulfate solution

c[(NH₄)₂ Fe(SO₄)₂] = 0.1 mol/l (0.1 N), concentrate for 250 mL titration solution, Titrisol®

Synonym(s):

Ammonium Ferrous Sulfate Solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352302
NACRES:
NA.21

product name

Ammonium iron(II) sulfate solution, for 250 mlc[(NH₄)₂ Fe(SO₄)₂] = 0.1 mol/l (0.1 N) Titrisol®

Quality Level

product line

Titrisol®

form

liquid

reaction suitability

reaction type: Complexometric reactions

concentration

0.1 M

technique(s)

titration: suitable

pH

0.4 (20 °C in H2O)

density

1.25 g/cm3 at 20 °C

storage temp.

15-25°C

Application


  • Regulation of Iron-Ion Transporter SLC11A2 by Three Identical miRNAs.: This study investigates the modulation of iron ion transporters, providing insights into molecular processes involving ammonium iron(II) sulfate, crucial for analytical chemistry applications (Sugino et al., 2022).

  • Two-Dimensional Bis(dithiolene)iron(II) Self-Powered UV Photodetectors with Ultrahigh Air Stability.: This research highlights the development of robust UV photodetectors using iron(II)-based materials, demonstrating the potential of ammonium iron(II) sulfate in creating high-performance electronic devices (Wang YC et al., 2021).

  • Pencil it in: pencil drawn electrochemical sensing platforms.: Focuses on innovative, cost-effective electrochemical sensors where iron plays a critical role, pertinent to applications in chemical analysis (Foster CW et al., 2016).

  • Catalytic effect of different forms of iron in purification of single-walled carbon nanotubes.: Discusses the use of various iron forms, including ammonium iron(II) sulfate, in enhancing the purification processes for carbon-based materials, significant in analytical applications (Suzuki T et al., 2010).

Legal Information

Titrisol is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service