Skip to Content
MilliporeSigma
All Photos(1)

Documents

1.09092

Supelco

Cerium(IV) sulfate solution

c(Ce(SO₄)₂ * 4 H₂O) = 0.1 mol/l (0.1 N), reag. Ph. Eur., ready-to-use volumetric solution for titration, Titriplex®

Synonym(s):

Cer sulfate, Cer(IV) sulfate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352302
NACRES:
NA.21

product name

Cerium(IV) sulfate solution, c(Ce(SO4)2 * 4 H2O) = 0.1 mol/l (0.1 N), Titripur®, reag. Ph. Eur.

Agency

reag. Ph. Eur.

Quality Level

product line

Titripur®

form

liquid

quality

Analyzed in our ISO 17025 accredited QC lab

reaction suitability

reaction type: Redox Reactions

concentration

0.1 M

technique(s)

titration: suitable

pH

0.4 (20 °C in H2O)

density

1.06 g/cm3 at 20 °C

storage temp.

15-25°C

Application


  • Humic acid removal using cellulose acetate membranes grafted with poly (methyl methacrylate) and aminated using tetraethylenepentamine.: This study highlights the application of cellulose acetate membranes modified with polymers for the efficient removal of humic acids from water, employing Cerium(IV) sulfate as a catalyst to initiate the polymerization process, which significantly enhances the membrane′s performance in water treatment (Gebru KA et al., 2018).

  • Characterization and ecological risk assessment of nanoparticulate CeO2 as a diesel fuel catalyst.: This research provides a detailed characterization of Cerium(IV) sulfate used as a precursor for nanoparticulate CeO2, assessing its ecological risks when applied as a catalyst in diesel fuels, offering insights into its environmental impact (Batley GE et al., 2013).

  • Chemiluminescence from an oxidation reaction of rhodamine B with cerium(IV) in a reversed micellar medium of cetyltrimethylammonium chloride in 1-hexanol-cyclohexane/water.: Explores the enhanced chemiluminescence reactions of Cerium(IV) sulfate, demonstrating its role in improving the sensitivity and specificity of analytical methods for detecting organic compounds (Hasanin TH et al., 2011).

  • Chemiluminescence reactions with cationic, neutral, and anionic ruthenium(II) complexes containing 2,2′-bipyridine and bathophenanthroline disulfonate ligands.: Investigates the chemiluminescence properties of various ruthenium(II) complexes, where Cerium(IV) sulfate serves as an effective oxidizing agent, highlighting its potential in analytical applications for detecting and measuring trace amounts of biological and chemical substances (Francis PS et al., 2010).

  • Chemiluminescence method for the determination of piroxicam by the enhancement of the tris-(4,7-diphenyl-1,10-phenanthrolinedisulphonic acid) ruthenium(II) (RuBPS)-cerium(IV) system and its application.: Presents a novel chemiluminescence technique for determining piroxicam, utilizing the catalytic properties of Cerium(IV) sulfate to enhance the luminescence signal, thereby offering a sensitive analytical tool for pharmaceutical analysis (Yu F et al., 2009).

Features and Benefits

Accreditation:
This volumetric solution is analyzed by our calibration laboratory D-K-15185-01-00 which is accredited according to DIN EN ISO/IEC 17025 for analysis of amount-of-substance concentrations in volumetric solutions by DAkkS (Deutsche Akkreditierungsstelle - German National Accreditation Body). The accreditation certificate can be found at www.sigmaaldrich.com/ISO17025.

Packaging

SKU-pack size numbers ending with a "3" are SmartChemicals with an RFID tag on the label for seamless data transfer to the instrument, e.g. xxxxxx1003 or xxxxxxx 4003. More information on www.sigmaaldrich.com/SmartChemicals

Linkage

Replaces: CX0612-6

Analysis Note

Form liquid
Amount-of-substance concentration 0.0995 - 0.1005 mol/L
Measurement uncertainty ± 0.0003 mol/L
Traceability NIST SRM

The concentration is determined by volumetric titration and refers to 20°C.
The amount-of-substance concentration of this volumetric solution is traceable to a primary standard reference material (SRM) from the National Institute of Standards and Technology, Gaithersburg, USA (NIST SRM 8040 di-sodium oxalate) by means of volumetric standard Iron(II)ethylendiammonium sulfate (article number 1.02402), certified reference material according to ISO 17034, analyzed by our accredited calibration laboratory of Merck KGaA, Darmstadt, Germany according to DIN EN ISO/IEC 17025. The uncertainty is expressed as expanded measurement uncertainty with a coverage factor k=2 covering a confidence level of 95%.

Note: The titer is a correction factor to correct for variations of the volumetric solution, the titration equipment, the temperature and other laboratory conditions. For correct titration results it is recommended to determine a titer with the laboratory specific equipment and under laboratory specific conditions directly after opening a new bottle and at regular time intervals.

Legal Information

Titripur is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Application Note details preparation of peracetic acid standard solution for analytical quality assurance (AQA) in testing laboratories.

Application Note details preparation of peracetic acid standard solution for analytical quality assurance (AQA) in testing laboratories.

Application Note details preparation of peracetic acid standard solution for analytical quality assurance (AQA) in testing laboratories.

Application Note details preparation of peracetic acid standard solution for analytical quality assurance (AQA) in testing laboratories.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service