Skip to Content
MilliporeSigma
All Photos(1)

Documents

1.03792

Supelco

Ammonium iron(II) sulfate hexahydrate

for analysis EMSURE® ISO

Synonym(s):

Ammonium iron(II) sulfate hexahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
H8FeN2O8S2 · 6H2O
CAS Number:
Molecular Weight:
392.14
MDL number:
UNSPSC Code:
12352302
EC Index Number:
233-151-8
NACRES:
NA.21

Agency

reag. ISO

Quality Level

product line

EMSURE®

Assay

99.0-101.5% (manganometric)

form

solid

potency

3250 mg/kg LD50, oral (Rat)

pH

3-5 (20 °C, 50 g/L in H2O)

mp

39-41 °C (decomposition)

solubility

269 g/L

density

1.86 g/cm3 at 20 °C

bulk density

900 kg/m3

anion traces

chloride (Cl-): ≤0.001%
phosphate (PO43-): ≤0.002%

cation traces

Ca: ≤0.002%
Cu: ≤0.002%
Fe3+: ≤0.02%
K: ≤0.01%
Mg: ≤0.01%
Mn: ≤0.05%
Na: ≤0.01%
Pb: ≤0.001%
Zn: ≤0.003%

storage temp.

15-25°C

InChI

1S/Fe.2H3N.2H2O4S.6H2O/c;;;2*1-5(2,3)4;;;;;;/h;2*1H3;2*(H2,1,2,3,4);6*1H2/q+3;;;;;;;;;;/p-2

InChI key

SHHFGSWBFZVUNE-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application


  • Water-Mediated Green and Efficient Synthesis of Bis(Indolyl)methanes Using Ammonium Iron(II) Sulfate.: This research highlights the use of ammonium iron(II) sulfate hexahydrate in the green synthesis of bis(indolyl)methanes. The study demonstrates the compound′s efficiency and eco-friendliness in catalyzing reactions in water, offering a sustainable alternative in organic synthesis (Letters in Organic Chemistry, 2020).


  • Determination of Ferromanganese and Ferrosilicon by Ammonium Ferrous Sulfate Titration and ICP.: This paper discusses the analytical methods for determining ferromanganese and ferrosilicon using ammonium iron(II) sulfate titration and inductively coupled plasma (ICP). The study underscores the compound′s role in precise quantitative analysis in metallurgical processes (Modern Analytical Chemistry Research, 2020).



Analysis Note

Assay (manganometric): 99.0 - 101.5 %
pH-value (5 %; water): 3.0 - 5.0
Chloride (Cl): ≤ 0.001 %
Phosphate (PO₄): ≤ 0.002 %
Iron(III)-salts (Fe³⁺): ≤ 0.02 %
Ca (Calcium): ≤ 0.002 %
Cu (Copper): ≤ 0.002 %
K (Potassium): ≤ 0.01 %
Mg (Magnesium): ≤ 0.01 %
Mn (Manganese): ≤ 0.05 %
Na (Sodium): ≤ 0.01 %
Pb (Lead): ≤ 0.001 %
Zn (Zinc): ≤ 0.003 %
Correspons to ISO

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service