Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

11721933001

Roche

DNA Molecular Weight Marker XIV (100 bp ladder)

solution, pkg of 50 μg (in 200 μl)

Synonym(s):

DNA marker

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105335

form

solution

Quality Level

packaging

pkg of 50 μg (in 200 μl)

manufacturer/tradename

Roche

concentration

250 μg/mL

color

colorless

solubility

water: miscible

storage temp.

−20°C

Related Categories

General description

Fragment mixture prepared by cleavage of a specially constructed plasmid with restriction endonucleases.
Size Range: 100 to 1500 bp and an additional band of 2642 bp

Application

DNA Molecular Weight Marker XIV (100 bp ladder) allows accurate sizing of DNA fragments generated by PCR or restriction digest or RT-PCR, following separation on agarose gels. The DNA molecular weight marker can be used in conjunction with DNA Molecular Weight Marker XIII (50 bp ladder) for precise size determination.
Electrophoretic separation of this molecular weight marker results in a regular pattern. The fragments have 5′-protruding ends and can be labeled with radioactive nucleotides (e.g.,32P-dTTP or 32P-dGTP) by standard filling-in reactions. End-labeling reactions can be performed with a radioactive or nonradioactive dideoxynucleotide (e.g., DIG-11-ddUTP) and Terminal Transferase.
DNA Molecular Weight Marker XIV has been used in gel electrophoresis.

Sequence

The mixture contains 15 double-stranded DNA fragments with the following base pair lengths (1 base pair = 660 daltons): 100, 200, 300, 400, 500, 600, 700, 800, 900, 1000, 1100, 1200, 1300, 1400, 1500, and an additional band of 2642 bp. The 500-bp and 1000-bp banding pattern is two to three times brighter.
Note: Fragment lengths are derived from computer analysis of the DNA sequence. Depending on the size range of the marker, the smallest fragments will only be visible on overloaded gels.To make them easy to identify, the 500 and 1,000 bp bands are two to three times as bright as the other bands in the ladder.

Unit Definition

50 μg = 1A260 unit

Physical form

Ready-to-use solution, 250 μg/ml, in TE buffer (10 mM Tris-HCl, 1 mM EDTA, pH 8.0).

Other Notes

For life science research only. Not for use in diagnostic procedures.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mirco Fanelli et al.
Nature protocols, 6(12), 1905-1919 (2011-11-16)
Formalin-fixed, paraffin-embedded (FFPE) samples represent the gold standard for storage of pathology samples. Here we describe pathology tissue chromatin immunoprecipitation (PAT-ChIP), a technique for extraction and high-throughput analysis, by techniques such as ChIP-seq, of chromatin derived from FFPE samples. Technically

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service