Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

11721925001

Roche

DNA Molecular Weight Marker XIII (50 bp ladder)

pkg of 50 μg (250 μg/ml), solution

Synonym(s):

DNA marker

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105335

form

solution

Quality Level

packaging

pkg of 50 μg (250 μg/ml)

manufacturer/tradename

Roche

concentration

250 μg/mL

color

colorless

solubility

water: miscible

storage temp.

−20°C

General description

Fragment mixture prepared by cleavage of a specially constructed plasmid with restriction endonucleases.
Size Range: 50 to 750 bp and an additional band of 2642 bp

Application

DNA Molecular Weight Marker XIII (50 bp ladder) allows accurate sizing of DNA fragments generated by PCR or restriction digest, followed by separation on agarose gels. DNA Molecular Weight Marker XIII (50 bp ladder) can be used in conjunction with DNA Molecular Weight Marker XIV (100 bp ladder) for precise size determination.
The fragments have 5′-protruding ends and can be labeled with radioactive nucleotides (e.g.,32P-dTTP or 32P-dGTP) by standard filling-in reactions. End-labeling reactions can be performed with a radioactive or nonradioactive dideoxynucleotide (e.g., DIG-11-ddUTP) and Terminal Transferase.
DNA Molecular Weight Marker for agarose gels; simplifies accurate molecular weight determination of double-stranded DNA fragments generated by:
  • Restriction digests
  • PCR
  • RT-PCR

Sequence

The mixture contains 15 double-stranded DNA fragments with the following base pair lengths (1 base pair = 660 daltons): 50, 100, 150, 200, 250, 300, 350, 400, 450, 500, 550, 600, 650, 700, 750, and an additional band of 2642 bp.
Note
: Fragment lengths are derived from computer analysis of the DNA sequence. Depending on the size range of the marker, the smallest fragments will only be visible on overloaded gels. The 250- and 500-bp banding pattern is two to three times brighter.

Unit Definition

50 μg = 1A260 unit

Physical form

Ready-to-use solution, 250μg/ml, in TE buffer (10mM Tris-HCl, 1mM EDTA, pH 8.0).

Preparation Note

Storage conditions (working solution): 2 to 8 °C
Once thawed we recommend further storage at 2 to 8 °C.

Other Notes

For life science research only. Not for use in diagnostic procedures.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development of a polymerase chain reaction system for the detection of dog and cat meat in meat mixtures and animal feed.
Abdulmawjood, A., Schonenbrucher, H. and Bulte, M.
Journal of Food Science, 68, 1757-1761 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service