Skip to Content
MilliporeSigma
All Photos(1)

Documents

11209264001

Roche

DNA Molecular Weight Marker VII

pkg of 50 μg (in 200 μl), solution

Synonym(s):

DNA marker

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105335

form

solution

Quality Level

packaging

pkg of 50 μg (in 200 μl)

manufacturer/tradename

Roche

concentration

250 μg/mL

color

colorless

solubility

water: miscible

storage temp.

−20°C

Related Categories

General description

Contents
Ready-to-use solution, 250 μg/ml, in TE buffer (Tris-HCl, 1 mM EDTA, pH 8.0).
Fragment mixture prepared by cleavage of SPP1-DNA with restriction endonuclease EcoR I.
The product is also available as DNA Molecular Weight Marker VII, Digoxigenin-labeled.
Size Range: 0.081 to 8.57 kbp

Application

DNA Molecular Weight Marker for agarose gels; simplifies accurate molecular weight determination of double-stranded DNA fragments generated by:
  • Restriction digests
  • PCR
  • RT-PCR

Sequence

The mixture contains seventeen fragments with the following base pair lengths (1 base pair = 660 daltons): 81, 359, 492, 710, 718*, 992, 1164, 1482, 1515, 1882, 1953, 2799, 3639, 4899, 6106, 7427, and 8576 as determined by computer analysis of the SSP1 sequence.
Note: Fragment lengths are derived from computer analysis of the DNA sequence. Depending on the size range of the marker, the smallest fragments will only be visible on overloaded gels.
*Alonso, J.C. and Trautner, T.A., personal communication of unpublished results, the 718-bp fragment is present in non-stoichometric amounts.

Unit Definition

50 μg = 1A260 unit

Preparation Note

Storage conditions (working solution): 2 to 8 °C
Once thawed store at 2 to 8 °C.

Other Notes

For life science research only. Not for use in diagnostic procedures.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service