Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SYMBSVB01

Millipore

Steritest® holder

For sterile bags and Steridilutor® vent chamber, for use with Steritest® Symbio pump

Synonym(s):

Bags and vent mount

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151818
NACRES:
NB.79

material

316L stainless steel

Agency

EP (2.6.1)
JP (4.06)
USP (71)

packaging

pkg of 1 unit

manufacturer/tradename

Steritest®

input

pharmaceutical(s)

application(s)

pharmaceutical
sterility testing

compatibility

for use with Steritest® Symbio FLEX Pump Kit, 2 media (SYMBFLE01)
for use with Steritest® Symbio ISL Pump Kit, 2 media (SYMBISL01)
for use with Steritest® Symbio LFH Pump Kit (SYMBLFH01)
for use with Steritest® Symbio pump

shipped in

ambient

General description

Steritest® holder is an accessory designed to keep the Steridilutor® Vent Chamber above the liquid level when reconstituting powders and diluting liquids. This ensures pressure release, thus preventing vial leakage. Free your work bench by hanging the sterile collapsible bags on the holder hooks. The holder is compatible with all Steritest® pumps.

Application

Steritest® holder is used to hang bags on the Steritest® Symbio bottle holder during liquid sampling and the vent of the Steridilutor® expansion chamber (an accessory used to dissolve and/or dilute powder samples in small vials before filtration on a Steritest® device)

Legal Information

STERIDILUTOR is a registered trademark of Merck KGaA, Darmstadt, Germany
STERITEST is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service