Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SYMBCAN08

Millipore

Steritest® carrying trays

for transportation and incubation of media-filled canisters, for use with Steritest® NEO Devices, pkg of (8 trays per pack)

Synonym(s):

Canister holder; Canister transportation aid

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151818
eCl@ss:
32014001
NACRES:
NB.24

material

polyphenylsulfone (PPSU)

Agency

EP 2.6.1
JP 4.06
USP 71

manufacturer/tradename

Steritest®

packaging

pkg of (8 trays per pack)

input

sample type pharmaceutical(s)

application(s)

pharmaceutical
sample preparation
sterility testing

compatibility

for use with Steritest® NEO Devices
for use with Steritest® Symbio FLEX Pump Kit, 2 media (SYMBFLE01)
for use with Steritest® Symbio ISL Pump Kit, 2 media (SYMBISL01)
for use with Steritest® Symbio LFH Pump Kit (SYMBLFH01)
for use with Steritest® Symbio Pump
standard for VHP (Autoclavable)

shipped in

ambient

General description

Steritest® carrying trays are the accessories that can be used with Steritest® NEO devices. A single carrying tray secures up to 5 canisters in an upright position at the end of the sterility test, facilitating safe transport of the canisters from the test environment to the incubator. It enables the user to visually inspect all 5 canisters at once by gently swirling the carrying tray during intermediate and final readings of the sterility test. The combination of 4 Steritest® carrying trays and a Steritest® rack enables the user to safely transport 20 media-filled canisters at once. The Steritest® canisters carrying trays and Steritest® rack can remain in the incubator.

Application

Steritest® carrying trays are used to streamline your workflow and increase safety during laboratory filtration and sterility testing.

Legal Information

STERITEST is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service