Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SYMBDTC01

Millipore

Steritest® Symbio 2 media drain tray container

for use with any type of Steritest® Symbio pump, this accessory streamlines the workflow and increases safety during sterility testing

Synonym(s):

Outlet

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151818
NACRES:
NB.79

material

316L stainless steel container (drain tray)

Agency

EP (2.6.1)
JP (4.06)
USP (71)

packaging

pkg of 1 unit

manufacturer/tradename

Steritest®

input

pharmaceutical(s)

application(s)

pharmaceutical
sterility testing

compatibility

for use with any type of Steritest® Symbio pump

shipped in

ambient

General description

Steritest® Symbio plastic fluid collector for drain tray. This tray is compatible with any type of Steritest® Symbio pump.

Application

Accessories for Streamlining Your Workflow and Increasing Safety during sterility testing.

Legal Information

STERITEST is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service