Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SYMBSYS01

Millipore

Steritest® Syringe Support

Steritest® accessory to hold syringe, For streamlining workflow and increasing safety

Synonym(s):

Valve

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151818
eCl@ss:
32014001
NACRES:
NB.24

material

316L stainless steel
EPDM rubber (foam)
Nylon 66 (glass bead reinforced)
Nylon
PVC
acrylonitrile-butadiene-styrene (ABS)
anodized aluminum
brass (nickel-plated)
polyoxymethylene
polyphenylsulfone (PPSU)

Agency

EP 2.6.1
JP 4.06
USP 71

packaging

pkg of 1 kit

manufacturer/tradename

Steritest®

input

sample type pharmaceutical(s)

application(s)

pharmaceutical
sample preparation
sterility testing

compatibility

for use with Steritest® Symbio FLEX Pump Kit, 2 media (SYMBFLE01)
for use with Steritest® Symbio ISL Pump Kit, 2 media (SYMBISL01)
for use with Steritest® Symbio LFH Pump Kit (SYMBLFH01)
for use with Steritest® Symbio Pump
standard for VHP

shipped in

ambient

General description

The STERITEST® Syringe Support is an accessory designed to be used with the STERITEST® EZ device for prefilled syringes with needles. The Syringe support valve is connected to the STERITEST® pump through a dedicated port on the pump communication hub. The distribution of sterile fluid to dilute the contents of the syringe is controlled by an automatic valve, eliminating the necessity to turn the dilution bottle over between each syringe tested. The STERITEST® Syringe Support is compatible with all STERITEST® Equinox and STERITEST® Symbio pumps. For STERITEST® Equinox pumps, the standard bottle holder needs to be replaced by a STERITEST® Symbio complete bottle holder >(SYMBBTH01) including the removable bottle basket. This accessory is autoclavable (except for the removable automatic valve) and resistant to vaporized hydrogen peroxide (VHP) and peracetic acid decontamination cycles in isolators.

Application

The STERITEST® Syringe Support is an accessory used to hold the syringe, and aids in the safe handling of syringes with needles. This accessory streamlines the workflow and increases safety during laboratory filtration and sterility testing.

Components

NA

Legal Information

STERITEST is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service