Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SNAP2MB2

Millipore

SNAP id® 2.0 Protein Detection System

Midi and MultiBlot

Synonym(s):

Western blot device, immunodetection device

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116010
eCl@ss:
42029053
NACRES:
NB.22

Product Name

SNAP i.d. 2.0 Protein Detection System-Midi and MultiBlot (8.5 x 13.5 cm and 4.5 x 8.4 cm), Developed to meet the needs of our Western blotting customers, the SNAP i.d. 2.0 system produces blots of a very high quality. Unique vacuum-driven technology & a built-in flow distributor actively drive reagents through the membrane.

manufacturer/tradename

SNAP id®

technique(s)

western blot: suitable

compatibility

for use with Commercially available blocking reagents
for use with Luminata Western HRP Substrates
for use with Nitrocellulose
for use with PVDF (Immobilon membranes)
for use with blØk<TMSYMBOL></TMSYMBOL>-CH Buffer (cat. no. WBAVDCH01)
for use with blØk<TMSYMBOL></TMSYMBOL>-FL Buffer (cat. no. WBAVDFL01)
for use with blØk<TMSYMBOL></TMSYMBOL>-PO Buffer (cat. no. WBAVDP001)
for use with commercially available detection reagents

detection method

chemiluminescent
colorimetric
fluorometric

shipped in

ambient

storage temp.

room temp

General description

This system includes the SNAP i.d. 2.0 Base, a Midi Blot Holding Frame,a MultiBlot Holding Frame, two Antibody Collection Trays, vacuum tubing, Rolling Pad & Blot Roller, two Wetting Trays and a Quick Start Guide

Application

Developed to meet the needs of our Western blotting customers, the SNAP i.d. 2.0 system produces blots of a very high quality. Unique vacuum-driven technology & a built-in flow distributor actively drive reagents through the membrane.
For use on immunoblots.

Linkage

Replaces: WBAVDBASE

Legal Information

SNAP id is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

The SNAP i.d. 2.0 Protein Detection System is the second generation of the SNAP i.d. method for detecting immunoreactive proteins on Western blots.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service