Skip to Content
MilliporeSigma

MA700

mA700 Essential Power Supply

Synonym(s):

power supply

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NC.01
UNSPSC Code:
41105303
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


packaging

pkg of 1 ea

General description

High capacity power supply for electrophoresis and Western blotting

MA700-USmA700 Essential Power Supply, US plug
MA700-EUmA700 Essential Power Supply, Euro plug
MA700-UKmA700 Essential Power Supply, UK plug
MA700-NImA700 Essential Power Supply, Japan plug
MA700-ZHmA700 Essential Power Supply, China plug

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Application

  • Enhanced capacity for protein and DNA electrophoresis
  • Tank & semidry protein transfer
  • Programmable
  • Color, LCD display shows all parameters during operation
  • Compact, stackable design

  • Gel Electrophoresis
  • Immunoblotting (Western)
  • Western Blotting





Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library






Global Trade Item Number

SKUGTIN
MA700-US04054839735707
MA700-EU04054839735714
MA700-UK04054839735721
MA700-NI04054839735837
MA700-ZH04054839735844
YP0304054839735752
YP2204054839735622
YP6104054839735738
YP1204054839735615
YP1804054839735745