Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SNAP2BHMN0100

Millipore

SNAP id® 2.0 Mini Blot Holders

7.4 x 8.4 cm, horizontal, flat profile for compact space storage

Synonym(s):

SNAP id® consumables, Western blot, Western blot holders

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116010
eCl@ss:
42029053
NACRES:
NB.22

Product Name

SNAP id® 2.0 Mini Blot Holders (7.5 x 8.4 cm), The new Mini Blot Holders ensure the membrane is horizontal allowing uniform Antibody distribution. The flat profile alows for compact space allocation for storage, shipping & waste management.

Quality Level

manufacturer/tradename

SNAP id®

technique(s)

western blot: suitable

compatibility

for use with Commercially available blocking reagents
for use with Luminata Western HRP Substrates
for use with Nitrocellulose
for use with PVDF (Immobilon membranes)
for use with blØk<TMSYMBOL></TMSYMBOL>-CH Buffer (cat. no. WBAVDCH01)
for use with blØk<TMSYMBOL></TMSYMBOL>-FL Buffer (cat. no. WBAVDFL01)
for use with blØk<TMSYMBOL></TMSYMBOL>-PO Buffer (cat. no. WBAVDP001)
for use with commercially available detection reagents

detection method

chemiluminescent
colorimetric
fluorometric

shipped in

ambient

storage temp.

room temp

General description

The new Mini Blot Holders ensure the membrane is horizontal allowing uniform Antibody distribution. The flat profile alows for compact space allocation for storage, shipping and waste management.

Application

For use on immunoblots.

Legal Information

SNAP id is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

Western blot protocol details protein transfer from gels to nitrocellulose, crucial for immunoblotting procedures in research.

Western blot protocol details protein transfer from gels to nitrocellulose, crucial for immunoblotting procedures in research.

Western blot protocol details protein transfer from gels to nitrocellulose, crucial for immunoblotting procedures in research.

Western blot protocol details protein transfer from gels to nitrocellulose, crucial for immunoblotting procedures in research.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service