Skip to Content
MilliporeSigma

MGT

mPAGE® Mini Gel Tank

for electrophoresis of precast gels or hand cast polyacrylamide gels

Synonym(s):

mini gel tank

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NC.01
UNSPSC Code:
41105340
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


AC/DC input

100 watts (maximum power limit), 300 V (maximum voltage limit)

manufacturer/tradename

Millipore

packaging

1 ea of (for running 2 or 4 gels)

parameter

70 °C ambient temp. limit

technique(s)

SDS-PAGE: suitable, electrophoresis: suitable

L × W × H

17 cm × 13.5 cm × 15.5 cm, tank and lid

buffer volume

1265 mL , Primary and secondary core (4 gels), 790 mL , Primary core (2 gels)

tank L × W × H

17 cm × 13 cm × 15.5 cm

compatibility

for use with mPAGE®

  • Gels cast with mPage® Gel Caster
  • Other precast gels with cassette dimension 8 x 10 cm
  • mPAGE® Bis-Tris Precast Gels
  • mPAGE® Mini Wet Transfer System

General description

The mPAGE® Mini Gel Tank is used to perform high-resolution protein separation on handcast or precast polyacrylamide gels. The gel tank is designed for use with 8 cm x 10 cm mini gels and is compatible with gels cast on the mPAGE® Gel Caster and mPAGE® Precast Gels. The mPAGE® Mini Gel Tank is available in 2 formats for running up to 2 or 4 mini gels and is designed to prevent incorrect assembly and leakage.

Other Notes

2-gel tank includes
  • mPAGE® Tank Lid with Electrode Cables (1)
  • mPAGE® Replacement Tank (1)
  • mPAGE® Primary Electrode Core (1)
  • mPAGE® Buffer Dam (1)
  • mPAGE® Gel Releasers (5)
4-gel tank includes
  • mPAGE® Tank Lid with Electrode Cables (1)
  • mPAGE® Replacement Tank (1)
  • mPAGE® Primary Electrode Core (1)
  • mPAGE® Secondary Electrode Core (1)
  • mPAGE® Buffer Dam (1)
  • mPAGE® Gel Releasers (5)

Legal Information

MPAGE is a registered trademark of Merck KGaA, Darmstadt, Germany




Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library