Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MSNUPSD50

Millipore

Multiscreen HTS Filter Plate, Plasmid

Plasmid Plate

Synonym(s):

Montage Plasmid Miniprep

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104923
eCl@ss:
32039006
NACRES:
NB.15

material

polystyrene

Quality Level

sterility

non-sterile

manufacturer/tradename

MultiScreen®

parameter

50-250 μL sample volume (per well)

technique(s)

DNA purification: suitable

plate size

96 wells

pore size

pore sizeproprietary

shipped in

ambient

Application

Research Category
Nucleic acid

Physical form

Color Code: Clear

Other Notes

Automation Compatible: Yes

Legal Information

MULTISCREEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service