Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MZFBN0W

Millipore

MultiScreenHTS 384 well

Opaque/White, Non-Sterile, Glass Fiber/Polyester mesh (FB) Filter Plate, 1.0 / 0.65 µm pore size, Device material Barex®/TiO2, 50 plates

Synonym(s):

384-Well MultiScreenHTS, HTS Filter Plate, MultiScreen 384 Plate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104923
eCl@ss:
32039006
NACRES:
NB.22

material

Barex®
flat bottom wells
glass fiber membrane
mesh polyester membrane
titanium dioxide

Quality Level

sterility

non-sterile

product line

MultiScreen®

feature

lid

manufacturer/tradename

MultiScreen®

parameter

20-100 μL sample volume (per well)

technique(s)

DNA purification: suitable
activity assay: suitable (with precipitations)
ligand binding assay: suitable
receptor binding assay: suitable

H

14.4 mm

L

128 mm

W

85.5 mm

filtration area

0.09 cm2

plate size

384 wells

well maximum volume

100 μL

working volume

20-100 μL

pore size

0.65 μm pore size
1.0 μm pore size

application(s)

sample preparation

binding type

high binding surface

Application

MultiScreenHTS 384 well has been used in elongation of very-long-chain fatty acid 1 (ELOVL1) assay.

Features and Benefits

MultiScreenHTS 384 well filter plates has the following features and benefits:
  • Developed for high-throughput automated workstations
  • Strong side-walls enhance handling
  • Provided with large surfaces for bar code labeling
  • Wells are individually sealed to prevent contamination
  • Removable underdrain for easy access to filters

Legal Information

Barex is a registered trademark of INEOS Barex AG
MULTISCREEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Discovery and Optimization of Pyrazole Amides as Inhibitors of ELOVL1
Come J H, et al.
Journal of Medicinal Chemistry, 64(24), 17753-17776 (2021)

Articles

ELISpot assay provides qualitative and quantitative information on immune responses, visualizing multiple secretory products from single responding cells.

ELISpot assay provides qualitative and quantitative information on immune responses, visualizing multiple secretory products from single responding cells.

ELISpot assay provides qualitative and quantitative information on immune responses, visualizing multiple secretory products from single responding cells.

ELISpot assay provides qualitative and quantitative information on immune responses, visualizing multiple secretory products from single responding cells.

Related Content

MultiScreenHTS 384-well filter plate for receptor-ligand binding assays using Human muscarinic M1 GPCR.

MultiScreenHTS 384-well filter plate for receptor-ligand binding assays using Human muscarinic M1 GPCR.

MultiScreenHTS 384-well filter plate for receptor-ligand binding assays using Human muscarinic M1 GPCR.

MultiScreenHTS 384-well filter plate for receptor-ligand binding assays using Human muscarinic M1 GPCR.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service