Skip to Content
MilliporeSigma
All Photos(1)

Documents

CLS3504

Corning® FiltrEX 96 well filter plates

0.2 μm pore size non-treated surface white polystyrene plate, PVDF membrane (hydrophilic), round wells, round bottom white polystyrene , rectangular bottom white polystyrene wells non-sterile, case of 50, pack of 10

Synonym(s):

96 well filter plate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104900
NACRES:
NB.24

product name

Corning® FiltrEX 96 well filter plates, non-treated surface, round bottom white polystyrene, non-sterile, pore size 0.2 μm, PVDF membrane (hydrophilic), lid: no, case of 50

material

PVDF membrane (hydrophilic)
rectangular bottom white polystyrene wells
round bottom white polystyrene
round wells
white polystyrene plate

sterility

non-sterile

feature

lid: no
lid: no
skirt
plate format: 96 well filter plate, standard

packaging

pack of 10
case of 50
case of 50

manufacturer/tradename

Corning 3504

maximum volume

350 μL

size

96 wells

working volume

350 μL

color

white

pore size

0.2 μm

binding type

non-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

The Corning FiltrEX 96 well white filter plate with 0.2μm hydrophilic PVDF membrane was designed with your application needs in mind. The low-binding hydrophilic PVDF membrane filter plates (CLS3504, CLS3505) are suitable for protein kinase assays or for bead and resin based separation assays.

  • Meets the ANSI/SBS1-2004 through ANSI/SBS 4-2004 standard dimensions to ensure automation compatibility
  • Rigid sidewalls make the plate ideal for automation handling
  • Wide skirt easily accommodates bar codes
  • Individual filter discs are sealed in the wells by a patented over-molding process to ensure 100% integrity of each well
  • The design of the nozzle prevents cross-contamination while allowing the plate to rest on the lab bench
  • 350μL working volume
  • Nonsterile
Glass fiber filter plates (CLS3510, CLS3511) can be used for a variety of applications, including plasmid isolation, DNA purification, or receptor/ligand binding assays. The low-binding hydrophilic PVDF membrane filter plates (CLS3504, CLS3505) are suitable for protein kinase assays or for bead and resin based separation assays.

Legal Information

Corning is a registered trademark of Corning, Inc.
FiltrEX is a trademark of Corning, Inc.

accessory


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service