Skip to Content
MilliporeSigma

M8IPS4510

Multiscreen® 96 well Plate, hydrophobic PVDF membrane

pore size 0.45 μm

Synonym(s):

Hydrophobic 96-Well Plate, PVDF 96-Well Plate, PVDF Membrane Plate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41104923
NACRES:
NB.22
eCl@ss:
32039006
Material:
flat bottom wells, PVDF membrane, clear acrylic plate
Pore size:
0.45 μm pore size
Sterility:
ethylene oxide treated
Binding type:
high binding surface
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

material

flat bottom wells, PVDF membrane, clear acrylic plate

description

Clear 96-well filter plate with 0.45 um pore size Hydrophobic PVDF membrane for Avidin-biotin Linkages, DNA-binding Proteins, Affects Function, ELISPOT Assays & Protein Interaction Assays.

sterility

ethylene oxide treated

usage

Automated

feature

hydrophobic, lid

packaging

pkg of 10 plates

manufacturer/tradename

MultiScreen®

technique(s)

ELISA: suitable, ELISPOT: suitable, activity assay: suitable (protein interaction), cell based assay: suitable

filtration area

0.28 cm2

plate size

96 wells

well maximum volume

300 μL

working volume

50-250 μL

matrix

Immobilon®-P

pore size

0.45 μm pore size

binding type

high binding surface

compatibility

centrifuge & manifold

shipped in

ambient

storage temp.

10-30°C

General description

Filter Code: IP
No matter what application you’re working with, Millipore has a plate developed to meet your specific needs. MultiScreen plates have a long history as a reliable tool for the advancement of drug discovery and life science research.

HTS Filter Plates (Plate catalogue numbers that start with MS)
- MultiScreenHTS plates are specifically developed for high-throughput use with automated work stations. Rigid sidewalls improve handling and ample surfaces are provided for bar code labels. Wells are individually sealed to prevent incubation crosstalk and the underdrain is removable for access to the filters. Filter plates are available in 96- and 384-well platforms with a broad selection of membranes and plastics.

- Millipore’s new MultiScreen HTS+ Hi flow filter plate design lessens non-specific binding and reduces variability in both background and signal intensities specifically for biochemical screening assays.

Classic Filter Plates (Plate catalogue numbers that start with MA)
- The original multiwell design is valuable for many applications which require immersing the filter in an assay tray. The easily removable underdrain and unique plate design are ideal for assays such as Elispot, PAMPA, cell screening (migration, invasion or chemotaxis) or for topcount detection users.

Accessory – MultiScreenHTS Vacuum Manifold
- The MultiScreenHTS Vacuum manifold was designed specifically to work with MultiScreenHTS filter plates. The combination results in optimal filtration characteristics and seamless automation integration.

Millicell Assay Plates
- In addition to the traditional assay plate format, filter plates are also available with cell culture trays for growing and assaying suspension and adherent cell lines. Cells grown on filter plates differentiate better than those grown on traditional plastic substrates, resulting in more in vivo-like results This filter plate/tray combination can be used for a variety of cellular assays, including, transport, toxicity and screening assays.

Application

Research Category
Nucleic acid

Protein

Assays

Radionucleotides

Physical form

Color Code: Clear

Other Notes

Automation Compatible: Yes

Legal Information

Immobilon is a registered trademark of Merck KGaA, Darmstadt, Germany
MULTISCREEN is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?




Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library