Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

MAB1998Z

Sigma-Aldrich

Anti-Integrin α2β1 Antibody, clone BHA2.1, azide free

clone BHA2.1, Chemicon®, from mouse

Synonym(s):

VLA-2, MAB1998

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

BHA2.1, monoclonal

species reactivity

human, pig

manufacturer/tradename

Chemicon®

technique(s)

flow cytometry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ITGA2(3673)

Related Categories

Specificity

Reacts with the collagen receptor VLA-2 (alpha2beta1) integrin complex.

Application

Research Category
Cell Structure
Research Sub Category
Integrins
This Anti-Integrin α2β1 Antibody, clone BHA2.1, azide free is validated for use in FC, IH, IP, FUNC for the detection of Integrin α2β1.
flow cytometry: 10 μg/mL

Immunoprecipitation: Immunoprecipitation of VLA-2 integrins from lysate of 10E6 cell equivalent requires 2-5 μg of BHA2.1

immunohistochemistry in frozen tissue sections: 5 μg/mL

Does not detect alpha2 integrin subunit by Western blot presumably due to conformational properties of the epitope involved. The epitope for BHA2.1 is dependent on the presence of the (I) domain of alpha2 integrin subunit and does not bind VLA-2 variant which lacks the I-domain.

Function Blocking: BHA2.1 inhibits VLA-2 mediated cell adhesion to collagen and laminin; complete inhibition can generally be achieved at a final concentration of 10 μg/mL.

Final working dilutions must be determined by end user.

Physical form

Format: Purified
Protein A purified
Purified immunoglobulin from Protein A chromatography. Liquid in 0.02M PB pH 7.6, 0.25M NaCl containing no preservative.

Storage and Stability

Maintain for 1 year at 2–8°C from date of shipment. Aliquot to avoid repeated freezing and thawing. For maximum recovery of product, centrifuge the original vial after thawing and prior to removing the cap.

Analysis Note

Control
Skin, CD4 & CD8 positive T cells

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Seiji Goda et al.
The American journal of pathology, 169(2), 445-458 (2006-08-01)
Natural killer (NK) cells play a key role in inflammation and tumor regression through their ability to migrate into tissues. CXCL12 is a chemokine that promotes lymphocyte invasion and migration into tissues; however, the mechanism for this process remains incompletely
Kwong-Fai Wong et al.
Oncotarget, 7(47), 77683-77695 (2016-10-22)
The Hippo pathway regulates the down-stream target Yes-associated protein (YAP) to maintain organ homeostasis, which is commonly inactivated in many types of cancers. However, how cell adhesion dysregulates the Hippo pathway activating YAP oncogene in hepatocellular carcinoma (HCC) remains unclear.
Christopher F Cummings et al.
The Journal of cell biology, 213(4), 479-494 (2016-05-25)
Basement membranes are defining features of the cellular microenvironment; however, little is known regarding their assembly outside cells. We report that extracellular Cl(-) ions signal the assembly of collagen IV networks outside cells by triggering a conformational switch within collagen
Josh Mauney et al.
Matrix biology : journal of the International Society for Matrix Biology, 28(5), 251-262 (2009-04-21)
Previously, we reported that the conformational transition of collagen I matrix plays, along with differentiation stimuli, a regulatory role in determination of differentiation lineage of bone marrow stromal sells via distinct signaling pathways specific for the structural state of the
Pei Jin Lim et al.
Frontiers in endocrinology, 14, 1195704-1195704 (2023-06-12)
Osteogenesis imperfecta (OI) is a heritable and chronically debilitating skeletal dysplasia. Patients with OI typically present with reduced bone mass, tendency for recurrent fractures, short stature and bowing deformities of the long bones. Mutations causative of OI have been identified

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service