Skip to Content
MilliporeSigma
All Photos(1)

Documents

MABT821

Sigma-Aldrich

Anti-Integrin beta1 (CD29) Antibody, clone mAb13

clone mAb13, from rat

Synonym(s):

Fibronectin receptor subunit beta, Glycoprotein IIa, GPIIA, VLA-4 subunit beta, CD29

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rat

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

mAb13, monoclonal

species reactivity

human

packaging

antibody small pack of 25 μL

technique(s)

immunohistochemistry: suitable
immunoprecipitation (IP): suitable
inhibition assay: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... ITGB1(3688)

General description

Integrin beta-1 (UniProt: P05556; also known as Fibronectin receptor subunit beta, Glycoprotein IIa, GPIIA, VLA-4 subunit beta, CD29) is encoded by the ITGB1 (also known as FNRB, MDF2, MSK12) gene (Gene ID: 3688) in human. Integrin beta-1 is a single-pass type I membrane protein that can associate with one of the multiple alpha subunits to serve as a receptor for fibronectin and other extracellular matrix proteins. Integrin beta-1 integrin recognizes the sequence R-G-D in a wide array of ligands. Five isoforms of integrin beta-1 have been reported that are produced by alternative splicing. Integrin beta-1 is synthesized with a signal peptide (aa 1-20) that is subsequently cleaved off. The mature form has an extracellular domain (aa 21-728), a helical domain (aa 729-751) and a cytoplasmic region (aa 752-798). Isoform 1 of integrin beta-1 is widely expressed and other isoforms are generally co-expressed with a more restricted distribution. Isoform 2 is expressed in skin, liver, skeletal muscle, cardiac muscle, placenta, umbilical vein endothelial cells, neuroblastoma cells, lymphoma cells, hepatoma cells and astrocytoma cells. Isoform 3 and isoform 4 are expressed in muscle, kidney, liver, placenta, cervical epithelium, umbilical vein endothelial cells, fibroblast cells, embryonal kidney cells, platelets and several blood cell lines. Isoform 4 is selectively expressed in peripheral T-cells and isoform 5 is expressed specifically in striated muscle (skeletal and cardiac muscle). Isoform 2 is reported to interfere with isoform 1 resulting in a dominant negative effect on cell adhesion and migration (in vitro).

Specificity

Clone mAb13 specifically detects beta 1 subunit of human integrin. It targets an epitope within 12 amino acids from the N-terminal half.

Immunogen

Fibronectin receptor (alpha V beta 1) purified from human placenta.

Application

Anti-Integrin beta1 (CD29), clone mAb13, Cat. No. MABT821, is a rat monoclonal antibody that detects integrin beta-1 and has been tested for use in Immunocytochemistry, Immunoprecipitation, and Inhibition studies.
Inhibition Analysis: A representative lot blocked cell attachment in Inhibition studies (Akiyama, S.K., et. al. (1989). J Cell Biol. 109(2):863-75; Humphries, J.D., et. al. (2005). J Biol Chem. 280(11):10234-43).

Immunocytochemistry Analysis: A representative lot detected Integrin beta1 (CD29) in Immunocytochemistry applications (Akiyama, S.K., et. al. (1989). J Cell Biol. 109(2):863-75; Humphries, J.D., et. al. (2005). J Biol Chem. 280(11):10234-43).

Immunoprecipitation Analysis: A representative lot detected Integrin beta1 (CD29) in Immunoprecipitation applications (Akiyama, S.K., et. al. (1989). J Cell Biol. 109(2):863-75).

Inhibition Analysis: A representative lot blocked HT-1080 cells adhesion to fibronectin coated plates (Courtesy of Dr. Kenneth M. Yamada, M.D., Ph.D., NIDCR, National Institutes of Health, Bethesda, MD USA).
Research Category
Cell Structure

Quality

Evaluated by Immunocytochemistry in A431 cells.

Immunocytochemistry Analysis: A 1:500 dilution of this antibody detected Integrin beta1 (CD29) in A431 cells.

Target description

88.42 kDa calculated.

Physical form

Format: Purified
Protein G purified
Purified rat monoclonal antibody IgG2a in PBS without preservatives.

Storage and Stability

Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Takuya Kuwashiro et al.
International journal of molecular medicine, 42(2), 957-965 (2018-05-23)
Although interferon (IFN)‑based treatment of patients with chronic hepatitis C virus (HCV) infection is widely applied, treatment resistance is often observed in patients with advanced liver fibrosis. Given that the molecular mechanisms of IFN resistance in liver fibrosis remain elusive, the
Sydney E Greer et al.
Molecular biology of the cell, 35(4), ar49-ar49 (2024-01-31)
Spatial and temporal regulation of chondrocyte maturation in the growth plate drives growth of many bones. One essential event to generate the ordered cell array characterizing growth plate cartilage is the formation of chondrocyte columns in the proliferative zone via
Mattia Vitale et al.
Gels (Basel, Switzerland), 8(5) (2022-05-28)
Hydrogel biomaterials mimic the natural extracellular matrix through their nanofibrous ultrastructure and composition and provide an appropriate environment for cell-matrix and cell-cell interactions within their polymeric network. Hydrogels can be modified with different proteins, cytokines, or cell-adhesion motifs to control
Md Saimon Mia et al.
European journal of cell biology, 102(2), 151316-151316 (2023-04-22)
The expression of the angiotensin-converting enzyme 2 (ACE2) is altered in multiple chronic kidney diseases like hypertension and renal fibrosis, where the signaling from the basal membrane proteins is critical for the development and progression of the various pathologies. Integrins
Ayan T Ray et al.
Development (Cambridge, England), 150(6) (2023-03-03)
Loss of FGF signaling leads to defects in salivary gland branching, but the mechanisms underlying this phenotype remain largely unknown. We disrupted expression of Fgfr1 and Fgfr2 in salivary gland epithelial cells and found that both receptors function coordinately in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service